Skip to Content
Merck
All Photos(1)

Documents

94029

Millipore

Legionella Selective Supplement IV

suitable for microbiology

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41171614
NACRES:
NA.74

Agency

according to ISO 11731:2017

Quality Level

form

powder

shelf life

limited shelf life, expiry date on the label

application(s)

environmental
food and beverages

microbiology

storage temp.

2-8°C

suitability

Legionella spp.

Application

An antibiotic supplement recommended for the selective isolation of Legionella species.

Components

(per vial, sufficient for 500 mL medium)
Polymyxin B sulfate 25′000 units,
Glycine 1.5 g,
Anisomycin 40mg,
Vancomycin 0.5mg,
Bromothymol blue 5mg,
Bromocresol purple 5mg

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service