Skip to Content
Merck
All Photos(1)

Key Documents

939056

Sigma-Aldrich

Sodium phosphate ChemBeads

new

Synonym(s):

Sodium orthophosphate ChemBeads, Trisodium phosphate ChemBeads

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Na3PO4
Molecular Weight:
163.94
UNSPSC Code:
12352100

description

Inorganic salt

Quality Level

form

solid

composition

14-16 wt% loading of base

reaction suitability

core: sodium

SMILES string

O=P(OO[Na])=O.[Na][Na]

InChI

1S/3Na.H3O4P/c;;;1-5(2,3)4/h;;;(H3,1,2,3,4)/q3*+1;/p-3

InChI key

RYFMWSXOAZQYPI-UHFFFAOYSA-K

General description

Sodium phosphate is a colorless to white crystalline powder or granules. It is prepared by neutralization of phosphoric acid under controlled conditions with sodium hydroxide or sodium carbonate .

Application

  • Sodium phosphate is used as a:
  • component in the preparation of solid crystals of calcium hydroxyapatite substituted by strontiumblock buffer to study segmental and subcellular distribution of CFTR in the kidney.
  • Other general uses are as pH control agent, water hardness precipitation, laboratory reagent, phosphatizing reagent.
For general uses, product is also available in powdered form (342483)

Features and Benefits

ChemBeads are chemical coated glass beads. ChemBeads offer improved flowability and chemical uniformity perfect for automated solid dispensing and high-throughput experimentation. The method of creating ChemBeads uses no other chemicals or surfactants allowing the user to accurately dispense sub-milligram amounts of chemical.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Unique Organization of Extracellular Amylases into Amylosomes in the Resistant Starch-Utilizing Human Colonic Firmicutes Bacterium Ruminococcus bromii
Ze X, et al.
mBio, 6(5), e01058-e01015 (2015)
The E3 ubiquitin ligase Trim7 mediates c-Jun/AP-1 activation by Ras signalling
Chakraborty A, et al.
Nature Communications, 6, 6782-6782 (2015)
Efficient reactivation of p53 in cancer cells by a dual MdmX/Mdm2 inhibitor
Qin L, et al.
Journal of the American Chemical Society, 136(52), 18023-18033 (2014)
ChemBead Enabled High-Throughput Cross-Electrophile Coupling Reveals a New Complementary Ligand
Aguirre A L, et al.
Chemistry (Weinheim An Der Bergstrasse, Germany), 27(51), 12981-12986 (2021)
Experience-dependent DNA methylation regulates plasticity in the developing visual cortex
Tognini P, et al.
Nature Neuroscience, 18(7), 956-958 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service