Skip to Content
Merck
All Photos(1)

Key Documents

900688

Sigma-Aldrich

Methanol

≥99.92%, suitable for UHPLC, mass spectrometry (MS)

Synonym(s):

Methyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OH
CAS Number:
Molecular Weight:
32.04
Beilstein:
1098229
EC Number:
MDL number:
UNSPSC Code:
12191601
NACRES:
NA.05
grade:
HPLC grade
technique(s):
HPLC: suitable
LC/MS: suitable
UHPLC: suitable
mass spectrometry (MS): suitable
application(s):
PFAS testing
food and beverages
bp:
64.7 °C (lit.)
vapor pressure:
410 mmHg ( 50 °C)
97.68 mmHg ( 20 °C)

Product Name

Methanol, UHPLC, suitable for mass spectrometry (MS)

grade

HPLC grade

Quality Level

Agency

suitable for ASTM® 7968
suitable for ASTM® 7979
suitable for DIN 38407-42
suitable for EPA 1621
suitable for EPA 1633
suitable for EPA 533
suitable for EPA 537.1
suitable for EPA 8327
suitable for EPA ACB B21-02
suitable for EPA ACB B23-05b
suitable for EPA OTM-45
suitable for GB 31604.35-2016
suitable for GB 5009.253-2016
suitable for ISO 21675 2019
suitable for ISO 25101
suitable for ISO/CEN 15968-2010

reg. compliance

suitable for FDA C-010.02

vapor density

1.11 (vs air)

vapor pressure

410 mmHg ( 50 °C)
97.68 mmHg ( 20 °C)

Assay

≥99.92%

form

liquid

autoignition temp.

725 °F

expl. lim.

36 %

technique(s)

HPLC: suitable
LC/MS: suitable
UHPLC: suitable
mass spectrometry (MS): suitable

refractive index

n20/D 1.329 (lit.)

bp

64.7 °C (lit.)

mp

−98 °C (lit.)

density

0.791 g/mL at 25 °C (lit.)

SMILES string

CO

application(s)

PFAS testing
food and beverages

format

neat

InChI

1S/CH4O/c1-2/h2H,1H3

InChI key

OKKJLVBELUTLKV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Methanol, a primary aliphatic alcohol, is an organic solvent that finds numerous uses in industrial applications. It is commonly used as an analytical standard, an extractant medium and as a mobile phase additive in various chromatographic separations.

Application

Methanol may be used:
  • As a stripping solvent to desorb pesticides after their extraction using hollow fiber liquid phase microextraction (HF-LPME), prior to the determination of the analytes in alcoholic beverages by ultra-high pressure liquid chromatography coupled to tandem mass spectrometry (UHPLC-MS/MS).
  • In the preparation of a mobile phase for the determination of phenolic acids in beverages by UPLC–MS/MS.
  • As an extraction solvent for the quantification of bisphenol-type endocrine disrupting compounds in food-contact recycled-paper materials by UPLC coupled to high resolution MS.

Legal Information

ASTM is a registered trademark of American Society for Testing and Materials

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rapid analysis of phenolic acids in beverages by UPLC-MS/MS
Gruz J, et al.
Food Chemistry, 111(3), 789-794 (2008)
David Pérez-Palacios et al.
Talanta, 99, 167-174 (2012-09-13)
Focused ultrasonic solid-liquid extraction (FUSLE) and reverse-phase ultra performance liquid chromatography (UPLC) coupled to a quadrupole-time of flight mass spectrometer (Q-TOF-MS) was applied to the determination of bisphenol-type endocrine disrupting compounds (EDCs) in food-contact recycled-paper materials. Recycled paper is a
Tae Joung Ha et al.
Food chemistry, 146, 270-277 (2013-11-02)
The present work was reported on investigation of saponin profiles in nine different legume seeds, including soybean, adzuki bean, cowpea, common bean, scarlet runner bean, lentil, chick pea, hyacinth bean, and broad bean using ultra performance liquid chromatography with photodiode
P Plaza Bolaños et al.
Journal of chromatography. A, 1208(1-2), 16-24 (2008-09-03)
An alternative method has been developed to determine more than 50 pesticides in alcoholic beverages using hollow fibre liquid phase microextraction (HF-LPME) followed by ultra-high pressure liquid chromatography coupled to tandem mass spectrometry (UHPLC-MS/MS), without any further clean-up step. Pesticides
Simultaneous gas chromatographic determination of methanol and free glycerol in biodiesel
Mittelbach M, et al.
Chromatographia, 42(7-8), 431-434 (1996)

Articles

A workflow for the sample preparation of hemp bud, hemp oil, chocolate, gummy, candy, and cream for the HPLC-PDA separation and analysis of 17 cannabinoids.

A workflow for the sample preparation of hemp bud, hemp oil, chocolate, gummy, candy, and cream for the HPLC-PDA separation and analysis of 17 cannabinoids.

Glyphosate analysis: LC-MS/MS method with Supel™ Carbon LC U/HPLC column for stability and retention.

Glyphosate analysis: LC-MS/MS method with Supel™ Carbon LC U/HPLC column for stability and retention.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service