Skip to Content
Merck

89922

Elemental Impurities according to ICH Q3D parenteral, Standard 2

TraceCERT®, (in 10% hydrochloric acid), applicable for testing acc. to USP<232>, Ph.Eur. Gen. Chapter 5.20

Synonym(s):

ICH Q3D Elemental Impurity Standard 2

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.24
UNSPSC Code:
41116107
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

agency

according to ICH Q3D, according to Ph. Eur., according to USP

product line

TraceCERT®

grade

certified reference material, TraceCERT®

shelf life

limited shelf life, expiry date on the label

composition

Au, 100 mg/L , Ir, 10 mg/L , Os, 10 mg/L , Pd, 10 mg/L , Pt, 10 mg/L , Rh, 10 mg/L , Ru, 10 mg/L

concentration

(in 10% hydrochloric acid)

technique(s)

ICP: suitable

format

multi-component solution

General description

This certified reference material (CRM) is produced in accordance with ISO 17034 and characterized in accordance with ISO/IEC 17025. This CRM is traceable to SI unit kg and measured against primary material from a National Metrology Institute (NMI), e.g. NIST.

Please visit ISO certificates and Site Quality Self-Assessments to access the current certificates of accreditation.

Search & download your certificate here.

Application

Elemental Impurities according to ICH Q3D parenteral can be used as calibration standard for quantitation of elemental impurities in tablets according to the new USP chapters <232> Elemental Impurities – Limits and <233> Elemental Impurities by ICP-OES method.

Preparation Note

For the preparation of this CRM only materials of highest purity were used.

Other Notes

We provide a diverse range of certified reference materials, including various elemental impurity mixes and single elements, with concentration limits that comply with ICH Q3D guidelines. For more information please visit our Inorganic Elemental Impurity Mix Standards.

In our effort to be more sustainable, we′re going digital with COAs-download them directly from our website.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany


pictograms

Corrosion

signalword

Warning

hcodes

Hazard Classifications

Met. Corr. 1

Storage Class

12 - Non Combustible Liquids



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library