Skip to Content

88554

Ethyl formate

analytical standard

Synonym(s):

Formic acid ethyl ester

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
HCOOC2H5
CAS Number:
Molecular Weight:
74.08
NACRES:
NA.24
PubChem Substance ID:
eCl@ss:
39022251
UNSPSC Code:
85151701
EC Number:
203-721-0
MDL number:
Beilstein/REAXYS Number:
906769
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

analytical standard

Quality Segment

vapor density

2.5 (vs air)

vapor pressure

15.16 psi ( 55 °C), 3.79 psi ( 20 °C)

assay

≥99.5% (GC)

autoignition temp.

851 °F

shelf life

limited shelf life, expiry date on the label

expl. lim.

16 %

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

refractive index

n20/D 1.359 (lit.), n20/D 1.360

bp

52-54 °C (lit.)

mp

−80 °C (lit.)

density

0.921 g/mL at 20 °C (lit.)

application(s)

agriculture
cleaning products
cosmetics
environmental
flavors and fragrances
food and beverages
personal care

format

neat

SMILES string

CCOC=O

InChI

1S/C3H6O2/c1-2-5-3-4/h3H,2H2,1H3

InChI key

WBJINCZRORDGAQ-UHFFFAOYSA-N

General description

Ethyl formate belongs to the group of ethyl esters, which can be synthesized via the reaction between formic acid and ethanol in the presence of sulfuric acid, under thermal conditions.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
Ethyl formate may be used as an analytical standard for the determination of the analyte in fruits, bread pre-ferment brew, alcoholic beverages, smog and gasoline samples by gas chromatography (GC) based techniques.

Other Notes

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.


Still not finding the right product?

Explore all of our products under Ethyl formate


pictograms

FlameExclamation mark

signalword

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

3 - Flammable liquids

wgk

WGK 1

flash_point_f

-4.0 °F - closed cup

flash_point_c

-20 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library