Skip to Content
Merck
All Photos(1)

Key Documents

85978

Supelco

Ethylene glycol

analytical standard

Synonym(s):

1,2-Ethanediol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOCH2CH2OH
CAS Number:
Molecular Weight:
62.07
Beilstein:
505945
EC Number:
MDL number:
UNSPSC Code:
41116107
eCl@ss:
39020501
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

analytical standard

Quality Level

vapor density

2.1 (vs air)

vapor pressure

0.08 mmHg ( 20 °C)

Assay

≥99.9% (GC)

autoignition temp.

752 °F

shelf life

limited shelf life, expiry date on the label

expl. lim.

15.3 %

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.431 (lit.)
n20/D 1.431

bp

195-198 °C

mp

−13 °C (lit.)

density

1.113 g/mL at 25 °C (lit.)

application(s)

environmental

format

neat

SMILES string

OCCO

InChI

1S/C2H6O2/c3-1-2-4/h3-4H,1-2H2

InChI key

LYCAIKOWRPUZTN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Ethylene glycol is a colorless liquid[1], which may be obtained via hydration of ethylene oxide.[2]
This substance is listed on the positive list of the EU regulation 10/2011 for plastics intended to come into contact with food.

Application

Ethylene glycol has been used as a solvent in the synthesis of silver nanostructures, which serve as effective catalysts in the epoxidation reaction of ethylene.[3]
It may be used as an analytical standard for the determination of the analyte in biological samples by various chromatography techniques.[4][5][6][7]
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - STOT RE 2 Oral

Target Organs

Kidney

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

239.0 °F - open cup

Flash Point(C)

115 °C - open cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

N M Wu et al.
Journal of analytical toxicology, 11(2), 63-66 (1987-03-01)
A simultaneous determination of ethylene glycol (EG) and ethylene chlorohydrin (EC), which are formed during sterilization of bone and dura mater with ethylene oxide, has been reported. The EG and EC residues in tissues were extracted with HPLC grade water
Petrochemical Processes null
Shape?and size?specific chemistry of Ag nanostructures in catalytic ethylene epoxidation
ChemCatChem, 2, 78-83 (2010)
H H Maurer et al.
Journal of chromatography. B, Biomedical sciences and applications, 754(2), 401-409 (2001-05-08)
A gas chromatographic-mass spectrometric assay is described for identification and quantification of the antifreezes ethylene glycol (EG) and diethylene glycol (DEG) in plasma for early diagnosis of a glycol intoxication. After addition of 1,3-propanediol as internal standard, the plasma sample
Alexandra Ehlers et al.
SpringerPlus, 2(1), 203-203 (2013-06-07)
A rapid headspace-gas chromatography (HS-GC) method was developed for the analysis of ethylene glycol and propylene glycol in plasma and serum specimens using 1,3-propanediol as the internal standard. The method employed a single-step derivitization using phenylboronic acid, was linear to

Questions

1–2 of 2 Questions  
  1. What is the Department of Transportation shipping information for this product?

    1 answer
    1. Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.

      Helpful?

  2. What is the molarity of ethylene glycol?

    1 answer
    1. Ethylene glycol is 17.93M.  This is calculated using the density (1.113 g/ml) and the molecular weight (62.07 g/mol).  If the product is less than 100% pure, correct for the purity.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service