Skip to Content
Merck
All Photos(1)

Documents

81503

Supelco

Poly(methyl methacrylate)

analytical standard, for GPC, 100,000

Synonym(s):

PMMA, Poly(methacrylic acid methyl ester)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[CH2C(CH3)(CO2CH3)]n
CAS Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard
for GPC

Quality Level

autoignition temp.

580 °F

shelf life

limited shelf life, expiry date on the label

mol wt

Mn ~91500
Mp ~102500
Mw ~98500

technique(s)

gel permeation chromatography (GPC): suitable

solubility

H2O: insoluble

density

1.200 g/cm3

Mw/Mn

~1.08

format

neat

storage temp.

2-8°C

InChI

1S/C5H9O2/c1-4(2)5(6)7-3/h1-3H3

InChI key

PMAMJWJDBDSDHV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Poly(methyl methacrylate) may be used as a reference suspension polymer for the analysis of the composition of poly(methyl methacrylate) (PMMA) by gas chromatography method.

Storage Class Code

11 - Combustible Solids

WGK

nwg

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Analysis of pyrolyzates of polystyrene and poly (methyl methacrylate) by gas chromatography.
Lehmann FA and Brauer GM
Analytical Chemistry, 33(6), 673-676 (1961)
Taher Alizadeh et al.
Journal of hazardous materials, 248-249, 401-406 (2013-02-19)
It was demonstrated that a thin film of chemically exfoliated graphene flakes, blended with poly(methyl methacrylate), could be used as a chemiresistor sensor for the sensitive and selective determination of formaldehyde vapor. Formaldehyde adsorption on the sensing film led to
A K Pathak et al.
The Journal of clinical pediatric dentistry, 37(3), 335-339 (2013-07-17)
Biofilms on removable orthodontic appliances act as reservoir of microorganisms, capable of modifying the environmental condition of oral cavity and are difficult to be removed with routine hygiene measures. The present investigation includes enumeration, identification and numerical analysis of different
Patrick M Honore et al.
ASAIO journal (American Society for Artificial Internal Organs : 1992), 59(2), 99-106 (2013-02-27)
In recent years, after all the attention has been focused on the dose for continuous renal replacement therapy (CRRT) in sepsis and systemic inflammation response syndrome (SIRS), the relatively negative results of all those studies did urge our expectations on
José F Alfonso et al.
Journal of refractive surgery (Thorofare, N.J. : 1995), 29(4), 266-272 (2013-04-06)
To assess the visual and refractive outcomes of implantation of intrastromal corneal ring segments (ICRS) in keratoconic eyes with no coincident topographic and comatic axes. Forty-one keratoconic eyes of 39 patients with no coincident topographic and comatic axes were implanted

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service