Skip to Content
Merck
All Photos(2)

Key Documents

79593

Supelco

(−)-Bilobalide

analytical standard

Synonym(s):

Bilobalide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H18O8
CAS Number:
Molecular Weight:
326.30
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥96.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

food and beverages

format

neat

storage temp.

2-8°C

SMILES string

CC(C)(C)[C@]1(O)C[C@@H]2OC(=O)C[C@]23C(=O)O[C@@H]4OC(=O)[C@H](O)C134

InChI

1S/C15H18O8/c1-12(2,3)14(20)4-6-13(5-7(16)21-6)10(19)23-11-15(13,14)8(17)9(18)22-11/h6,8,11,17,20H,4-5H2,1-3H3/t6-,8-,11-,13+,14+,15?/m0/s1

InChI key

MOLPUWBMSBJXER-DXPZGIEZSA-N

Looking for similar products? Visit Product Comparison Guide

General description

(−)-Bilobalide is a Ginkgo terpenoid, available as the main pharmacological component in Ginkgo biloba extract. It exhibits neuroprotectant activity.

Application

(−)-Bilobalide may be used as an analytical rstandard for the quantification of the analyte in biological samples using liquid chromatography coupled to tandem mass spectrometry.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

This compound is commonly found in plants of the genus: ginkgo

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

LC-ESI-MS determination of bilobalide and ginkgolides in canine plasma
Tang J, et al.
Chromatographia, 63(1-2), 53-58 (2006)
Simultaneous determination of ginkgolides A, B, C and bilobalide in plasma by LC-MS/MS and its application to the pharmacokinetic study of Ginkgo biloba extract in rats
Xie J, et al.
Journal of Chromatography. B, Biomedical Sciences and Applications, 864(1-2), 87-94 (2008)
T Yoshitake et al.
British journal of pharmacology, 159(3), 659-668 (2010-01-29)
Experimental and clinical data suggest that extracts of Ginkgo biloba improve cognitive function. However, the neurochemical correlates of these effects are not yet fully clarified. The purpose of this study was to examine the effects of acute and repeated oral
Karin Woelkart et al.
Phytotherapy research : PTR, 24(3), 445-450 (2009-12-31)
A sensitive LC-ESI-MS method with a solid-phase extraction was established for the determination of bilobalide, ginkgolide A and ginkgolide B in human plasma; bioavailability and pharmacokinetics of three different Ginkgo biloba L. preparations have been investigated. The preparations used in
Etsuko Suzuki et al.
The journal of physiological sciences : JPS, 61(5), 421-427 (2011-06-29)
Bilobalide, a unique constituent of Ginkgo biloba, has been reported to potentiate population spikes in hippocampal CA1 pyramidal cells and to protect the brain against cell death. In this study, the effects of bilobalide on synaptic transmission and its plasticity

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service