Skip to Content
Merck

78419

Salmonella ChromoSelect Agar

NutriSelect® Plus, suitable for microbiology

Synonym(s):

Salmonella Agar ChromoSelect

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
41171606
NACRES:
NA.74
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


sterility

non-sterile

Quality Segment

form

powder

shelf life

limited shelf life, expiry date on the label

composition

agar, 13 g/L , bile salt mixture, 1 g/L , chromogenic mixture, 5.4 g/L , peptic digest of animal tissue, 6 g/L , yeast extract, 2.5 g/L

manufacturer/tradename

NutriSelect® Plus

pH

7.7±0.2 (25 °C)

application(s)

environmental
food and beverages, microbiology

suitability

selective and differential for Salmonella spp., selective and differential for Shigella spp.

Application

Salmonella ChromoSelect Agar is a selective medium used for simultaneous detection of Escherichia coli and Salmonella from food and water. Escherichia coli and Salmonella are easily distinguishable due to the colony characteristics. Salmonella give light purple colonies a halo. Escherichia coli has a characteristic blue color, other organisms give colorless colonies. The characteristic light purple and blue color is due to the chromogenic mixture.

Preparation Note

Suspend 27.9 g in 1 litre distilled water. Boil gently to dissolve the medium completely. DO NOT AUTOCLAVE. Cool to 50°C. Mix well and pour into sterile petri plates.

Other Notes

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany


Storage Class

11 - Combustible Solids

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library