Skip to Content
Merck

77147

Color Reference Solution acc. to APHA

analytical standard, set of 20 × 10 mL 20 standard solutions

Synonym(s):

APHA color reference solution

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.24
UNSPSC Code:
41116107
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

analytical standard

Quality Segment

agency

according to APHA

shelf life

limited shelf life, expiry date on the label

packaging

set of 20 × 10 mL 20 standard solutions

application(s)

cleaning products
coatings
cosmetics
environmental
flavors and fragrances
food and beverages
glass & ceramic
personal care
pharmaceutical

format

multi-component solution

General description

The kit contains a set of 20 color reference solutions defined by ASTM D1209-00, spanning concentrations of platinum cobalt to water from 0 to 500 ppm.
contains Cobalt(II) chloride

Application

The color reference solution finds its use in routine calibration of colour measuring instruments and validation of test results.

Features and Benefits

  • Ready-to-use solutions
  • Compliant to ASTM D 1209-00
  • Available in tamper-evident 10 mL ampules
  • Expiry dated on the label with a limited shelf life
  • Accompanied with a detailed certificate of analysis for an effective quality management system

Packaging

ampules with 10 mL solution

Other Notes

This is a set of the following color reference solutions according to APHA:

  • Colour Reference Solution 5 - 10mL ampule
  • Colour Reference Solution 10 - 10mL ampule
  • Colour Reference Solution 15 - 10mL ampule
  • Colour Reference Solution 20 - 10mL ampule
  • Colour Reference Solution 25 - 10mL ampule
  • Colour Reference Solution 30 - 10mL ampule
  • Colour Reference Solution 35 - 10mL ampule
  • Colour Reference Solution 40 - 10mL ampule
  • Colour Reference Solution 50 - 10mL ampule
  • Colour Reference Solution 60 - 10mL ampule
  • Colour Reference Solution 70 - 10mL ampule
  • Colour Reference Solution 100 - 10mL ampule
  • Colour Reference Solution 150 - 10mL ampule
  • Colour Reference Solution 200 - 10mL ampule
  • Colour Reference Solution 250 - 10mL ampule
  • Colour Reference Solution 300 - 10mL ampule
  • Colour Reference Solution 350 - 10mL ampule
  • Colour Reference Solution 400 - 10mL ampule
  • Colour Reference Solution 450 - 10mL ampule
  • Colour Reference Solution 500 - 10mL ampule


Still not finding the right product?


signalword

Danger

Hazard Classifications

Aquatic Chronic 3 - Carc. 1B - Eye Irrit. 2 - Met. Corr. 1 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type ABEK (EN14387) respirator filter

wgk

WGK 2



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library