Skip to Content
Merck

76969

1-Pentene

analytical standard

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
CH3CH2CH2CH=CH2
CAS Number:
Molecular Weight:
70.13
UNSPSC Code:
41116107
NACRES:
NA.24
PubChem Substance ID:
EC Number:
203-694-5
Beilstein/REAXYS Number:
1731629
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

analytical standard

Quality Segment

vapor density

2.4 (vs air)

vapor pressure

10.27 psi ( 20 °C)

assay

≥99.5% (GC)

autoignition temp.

522 °F

shelf life

limited shelf life, expiry date on the label

expl. lim.

8.7 %

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

refractive index

n20/D 1.371 (lit.), n20/D 1.372

bp

29.9-30.1 °C (lit.)

density

0.641 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
environmental
food and beverages
personal care
petroleum

format

neat

storage temp.

2-8°C

SMILES string

CCCC=C

InChI

1S/C5H10/c1-3-5-4-2/h3H,1,4-5H2,2H3

InChI key

YWAKXRMUMFPDSH-UHFFFAOYSA-N

General description

This substance is listed on the positive list of the EU regulation 10/2011 for plastics intended to come into contact with food. Find all available reference materials for compounds listed in 10/2011 here

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.


Still not finding the right product?

Explore all of our products under 1-Pentene


signalword

Danger

Hazard Classifications

Asp. Tox. 1 - Eye Irrit. 2 - Flam. Liq. 1 - Skin Irrit. 2

Storage Class

3 - Flammable liquids

wgk

WGK 2

flash_point_f

-59.8 °F - closed cup

flash_point_c

-51 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library