Skip to Content
Merck
All Photos(2)

Documents

76514

Supelco

2,4-D

certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Synonym(s):

2,4-Dichlorophenoxyacetic acid, 2,4-D

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cl2C6H3OCH2CO2H
CAS Number:
Molecular Weight:
221.04
Beilstein:
1214242
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material
TraceCERT®

Quality Level

vapor pressure

0.4 mmHg ( 160 °C)

product line

TraceCERT®

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

136-140 °C (lit.)

format

neat

SMILES string

OC(=O)COc1ccc(Cl)cc1Cl

InChI

1S/C8H6Cl2O3/c9-5-1-2-7(6(10)3-5)13-4-8(11)12/h1-3H,4H2,(H,11,12)

InChI key

OVSKIKFHRZPJSS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.
Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 3 - Eye Dam. 1 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Manoharan Rajesh et al.
Protoplasma, 251(5), 1231-1243 (2014-03-19)
Podophyllum hexandrum Royle known as Indian mayapple is an important medicinal plant found only in higher altitudes (2,700 to 4,200 m) of the Himalayas. The highly valued anticancer drug Podophyllotoxin is obtained from the roots of this plant. Due to
Miguel Cacho Teixeira et al.
Trends in biotechnology, 25(8), 363-370 (2007-06-20)
Genomic information and tools are beginning to be used to increase our understanding of how organisms of all types interact with their environment. The study of the expression of all genes, at the genome, transcriptome, proteome and metabolome level, in
Heping Han et al.
Pest management science, 69(9), 996-1000 (2013-06-21)
The auxinic herbicide 2,4-D amine is known, in vitro, as a cytochrome P450 inducer. The current study uses 2,4-D pre-treatment, at the whole plant level, to study mechanism(s) of non-target site based herbicide resistance to the ACCase-inhibiting herbicide diclofop-methyl in
David H Garabrant et al.
Critical reviews in toxicology, 32(4), 233-257 (2002-08-20)
The scientific evidence in humans and animals relevant to cancer risks, neurologic disease, reproductive risks, and immunotoxicity of 2,4-D was reviewed. Despite several thorough in vitro and in vivo animal studies, no experimental evidence exists supporting the theory that 2,4-D
Lesa L Aylward et al.
Environmental health perspectives, 118(2), 177-181 (2010-02-04)
Several extensive studies of exposure to 2,4-dichlorophenoxyacetic acid (2,4-D) using urinary concentrations in samples from the general population, farm applicators, and farm family members are now available. Reference doses (RfDs) exist for 2,4-D, and Biomonitoring Equivalents (BEs; concentrations in urine

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service