Skip to Content
Merck
All Photos(2)

Key Documents

75541

Supelco

(−)-trans-Caryophyllene

analytical standard

Synonym(s):

β-Caryophyllene, trans-(1R,9S)-8-Methylene-4,11,11-trimethylbicyclo[7.2.0]undec-4-ene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H24
CAS Number:
Molecular Weight:
204.35
Beilstein:
2044564
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

analytical standard

Quality Level

Assay

≥98.0% (sum of enantiomers, GC)

optical activity

[α]20/D -10±1°, neat

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

bp

262-264 °C (lit.)

density

0.902 g/mL at 20 °C (lit.)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

storage temp.

2-8°C

SMILES string

[H][C@@]12CC\C(C)=C\CCC(=C)[C@H]1CC2(C)C

InChI

1S/C15H24/c1-11-6-5-7-12(2)13-10-15(3,4)14(13)9-8-11/h6,13-14H,2,5,7-10H2,1,3-4H3/b11-6+/t13-,14-/m1/s1

InChI key

NPNUFJAVOOONJE-GFUGXAQUSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

This compound is commonly found in plants of the genus: cinnamomum melissa

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Asp. Tox. 1 - Skin Sens. 1B

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

221.9 °F - closed cup

Flash Point(C)

105.5 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Berta Alquézar et al.
Scientific reports, 7(1), 5639-5639 (2017-07-19)
Production of citrus, the main fruit tree crop worldwide, is severely threatened by Huanglongbing (HLB), for which as yet a cure is not available. Spread of this bacterial disease in America and Asia is intimately connected with dispersal and feeding
Rajesh K Joshi
Natural product communications, 8(3), 401-402 (2013-05-18)
The essential oil composition from the aerial parts of Baccharoides lilacina (Dalzell & A. Gibson) M. R. Almeida was analyzed by gas chromatography (GC) and gas chromatography/mass spectrometry (GC/MS). A total of 41 compounds have been identified, representing 97.4% of
In-Young Choi et al.
The American journal of pathology, 182(3), 928-939 (2013-02-19)
The type 2 cannabinoid receptor (CB2R) was recently shown to mediate neuroprotection in ischemic injury. However, the role of CB2Rs in the central nervous system, especially neuronal and glial CB2Rs in the cortex, remains unclear. We, therefore, investigated anti-ischemic mechanisms
Jullyana de S S Quintans et al.
Planta medica, 79(2), 123-130 (2013-01-12)
Xylopia laevigata, popularly known as "meiú" and "pindaíba", is a medicinal plant used in the folk medicine of the Brazilian Northeast for several purposes. The chemical constituents of the essential oil from leaves of X. laevigata, collected from wild plants
Rajesh K Joshi
Natural product communications, 8(2), 225-226 (2013-03-22)
The hydro-distilled essential oil of dried flowers of Anaphalis contorta Hook f. has been examined by means of gas chromatography-mass spectrometry (GC-MS). The oil constituents were identified according to their mass spectra and their relative retention indices determined on a

Articles

A throughput HS-SPME-GC-MS method to analyze volatile compounds in Perilla leaves at low extraction temperatures using four different SPME fibers.

A throughput HS-SPME-GC-MS method to analyze volatile compounds in Perilla leaves at low extraction temperatures using four different SPME fibers.

A throughput HS-SPME-GC-MS method to analyze volatile compounds in Perilla leaves at low extraction temperatures using four different SPME fibers.

A throughput HS-SPME-GC-MS method to analyze volatile compounds in Perilla leaves at low extraction temperatures using four different SPME fibers.

Protocols

Fast GC analysis of sweet orange essential oil in hexane. Key components identified includes: β-Farnesene; α-Huµlene; Germacrene D; (+)-Valencene; Bicyclogermacrene; (+)-δ-Cadinene

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service