Skip to Content
Merck
All Photos(2)

Key Documents

75251

Supelco

Corilagin

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H22O18
CAS Number:
Molecular Weight:
634.45
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥98.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

food and beverages

format

neat

storage temp.

2-8°C

SMILES string

O[C@@H]1[C@H]2COC(=O)c3cc(O)c(O)c(O)c3-c4c(O)c(O)c(O)cc4C(=O)O[C@@H]1[C@@H](O)[C@@H](O2)OC(=O)c5cc(O)c(O)c(O)c5

InChI

1S/C27H22O18/c28-9-1-6(2-10(29)16(9)32)24(39)45-27-22(38)23-19(35)13(43-27)5-42-25(40)7-3-11(30)17(33)20(36)14(7)15-8(26(41)44-23)4-12(31)18(34)21(15)37/h1-4,13,19,22-23,27-38H,5H2/t13-,19-,22-,23+,27+/m1/s1

InChI key

TUSDEZXZIZRFGC-XIGLUPEJSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Biochem/physiol Actions

Corilagin is a polyphenol, specifically a hydrolyzable tannin, found in Traditional Chinese Medicine herbal preparations. It is reported to have anti-inflammatory and anti-tumor activities.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Christian Agyare et al.
Phytomedicine : international journal of phytotherapy and phytopharmacology, 18(7), 617-624 (2010-11-03)
Leaves from Phyllanthus muellerianus (Kuntze) Exell. are traditionally used for wound healing in Western Africa. Aqueous extracts of dried leaves recently have been shown to stimulate proliferation of human keratinocytes and dermal fibroblasts. Within bioassay-guided fractionation the ellagitannins geraniin (1)
Lei Zhao et al.
International immunopharmacology, 8(7), 1059-1064 (2008-05-20)
Corilagin (beta-1-O-galloyl-3,6-(R)-hexahydroxydiphenoyl-D-glucose) is a novel member of the tannin family which has been discovered from many medicinal plants and has been confirmed in many pharmacological activities. However, the purified Corilagin that was used in experiment is rare, and the anti-inflammatory
Bo Shen et al.
Hepatology (Baltimore, Md.), 47(2), 473-483 (2007-12-25)
Hepatic oxidative stress plays a critical role in metabolic forms of steatohepatitis. Phyllanthus urinaria, an herbal medicine, has been reported to have potential antioxidant properties. We tested the effects of P. urinaria on nutritional steatohepatitis both in vitro and in
Yuan-Jin Guo et al.
European journal of pharmacology, 635(1-3), 79-86 (2010-03-27)
The aim of this explore is to study the anti-inflammatory effect of Corilagin in herpes simplex virus (HSV)-1 infected microglial cells and HSV-1 infected mouse brain. The cellular model was set with microglial cells stimulated by HSV-1 and divided respectively
Weigang Duan et al.
Yakugaku zasshi : Journal of the Pharmaceutical Society of Japan, 125(7), 587-591 (2005-07-06)
Oxidized low-density lipoprotein (ox-LDL) is the main etiologic factor in atherogenesis, and antioxidants are accepted as effective treatment of atherosclerosis. The aim of this study was to clarify whether the mechanism of the antiatherogenic effects of the herb Phyllanthus Emblica

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service