Skip to Content
Merck

72004

dNTP Mix (10mM)

suitable for PCR, suitable for RT-PCR

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.55
UNSPSC Code:
41106301
Form:
liquid
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

liquid

Quality Segment

concentration

10 mM

technique(s)

PCR: suitable, RT-PCR: suitable

shipped in

dry ice

storage temp.

−20°C

General description

dNTP Mix (10mM) is an equimolar mixture consisting of high purity dATP, dGTP, dCTP, and dTTP which can be directly used in PCR, Reverse Transcription (RT) and one-step RT-PCR experiments.

Features and Benefits

dNTP mix has the following benefits:
  • HPLC purity >99%.
  • Strict Quality Control to ensure batch quality.



Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library


Related Content





Global Trade Item Number

SKUGTIN
72004-1ML04065267927510
72004-5ML04065267927527