71038BC
Assurance® GDS Cronobacter Tq II Kit
suitable for PCR testing in Food and Beverages applications
About This Item
Recommended Products
Product Name
GDS Cronobacter Tq II, BioControl Systems, for use with GDS Rotor-Gene®
Agency
according to AOAC® (PTM)
according to MicroVal
Quality Level
product line
Assurance GDS®
shelf life
6‑12 mo.
manufacturer/tradename
BioControl Systems
technique(s)
PCR: suitable
application(s)
food and beverages
pathogen testing
compatibility
for use with GDS Rotor-Gene®
storage temp.
2-8°C
suitability
Cronobacter spp. (spp.)
General description
Application
Features and Benefits
- Simple 1-step enrichment procedure for powdered infant formula, ingredients, and environmental samples
- Results are available in 22-26 hours, including enrichment, sample prep and analysis
Components
- Cronobacter Amplification Tubes Tq
- Cronobacter Concentration Reagant
- Resuspension Buffer
- Wash Solution
Compatibility
Testing Method
Legal Information
Storage Class Code
10 - Combustible liquids
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Articles
Assurance® GDS offers integrated solution for fast and accurate food pathogen detection in microbiology testing.
Assurance® GDS offers integrated solution for fast and accurate food pathogen detection in microbiology testing.
Assurance® GDS offers integrated solution for fast and accurate food pathogen detection in microbiology testing.
Assurance® GDS offers integrated solution for fast and accurate food pathogen detection in microbiology testing.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service