Skip to Content
Merck
All Photos(2)

Key Documents

69532

Supelco

Artemisinin

analytical standard

Synonym(s):

Arteannuin, Qinghaosu

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H22O5
CAS Number:
Molecular Weight:
282.33
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥98.0% (TLC)

optical activity

[α]/D +75.0±3°, c = 0.5 in methanol

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

156-157 °C (lit.)

application(s)

food and beverages

format

neat

SMILES string

C[C@@H]1CC[C@H]2[C@@H](C)C(=O)O[C@@H]3OC4(C)CC[C@@H]1[C@@]23OO4

InChI

1S/C15H22O5/c1-8-4-5-11-9(2)12(16)17-13-15(11)10(8)6-7-14(3,18-13)19-20-15/h8-11,13H,4-7H2,1-3H3/t8-,9-,10+,11+,13-,14-,15-/m1/s1

InChI key

BLUAFEHZUWYNDE-NNWCWBAJSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pamela J Weathers et al.
Journal of ethnopharmacology, 153(3), 732-736 (2014-03-26)
The Chinese have used Artemisia annua as a tea infusion to treat fever for >2000 years. The active component is artemisinin. Previously we showed that when compared to mice fed an equal amount of pure artemisinin, a single oral dose
C Wongsrichanalai et al.
Clinical microbiology and infection : the official publication of the European Society of Clinical Microbiology and Infectious Diseases, 19(10), 908-916 (2013-09-17)
Following a decade-long scale up of malaria control through vector control interventions, the introduction of rapid diagnostic tests and highly efficacious Artemisinin-based Combination Therapy (ACT) along with other measures, global malaria incidence declined significantly. The recent development of artemisinin resistance
Benjamin J Visser et al.
Expert opinion on pharmacotherapy, 15(15), 2219-2254 (2014-08-12)
Chemotherapy of malaria has become a rapidly changing field. Less than two decades ago, treatment regimens were increasingly bound to fail due to emerging drug resistance against 4-aminoquinolines and sulfa compounds. By now, artemisinin-based combination therapies (ACTs) constitute the standard
Victor M Ye et al.
Biotechnology journal, 7(1), 20-33 (2011-10-25)
Driven by requirements for sustainability as well as affordability and efficiency, metabolic engineering of plants and microorganisms is increasingly being pursued to produce compounds for clinical applications. This review discusses three such examples of the clinical relevance of metabolic engineering:
Moran Farhi et al.
Biotechnology & genetic engineering reviews, 29, 135-148 (2014-02-27)
Artemisinin, a natural compound from Artemisia annua, is highly effective in treating drug-resistant malaria. Because chemical synthesis of this natural terpenoid is not economically feasible, its only source remains as the native plant which produces only small quantities of it

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service