Skip to Content
Merck
All Photos(2)

Key Documents

68775

Supelco

Sunset Yellow FCF

analytical standard

Synonym(s):

Food Yellow 3, Orange Yellow S

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H10N2Na2O7S2
CAS Number:
Molecular Weight:
452.37
Colour Index Number:
15985
EC Number:
MDL number:
UNSPSC Code:
12164500
PubChem Substance ID:
E Number:
E110
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥95% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

>300 °C (lit.)

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

SMILES string

[Na+].[Na+].Oc1ccc2cc(ccc2c1\N=N\c3ccc(cc3)S([O-])(=O)=O)S([O-])(=O)=O

InChI

1S/C16H12N2O7S2.2Na/c19-15-8-1-10-9-13(27(23,24)25)6-7-14(10)16(15)18-17-11-2-4-12(5-3-11)26(20,21)22;;/h1-9,19H,(H,20,21,22)(H,23,24,25);;/q;2*+1/p-2/b18-17+;;

InChI key

OIQPTROHQCGFEF-QIKYXUGXSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Sunset Yellow FCF (E110) is a reddish-orange azo dye used as a synthetic colorant in various foods and beverages to improve their texture and appearance.

Application

The analytical standards can also be used as follows:
  • Simultaneous analysis of Sunset Yellow FCF, Allura Red AC, Quinoline Yellow WS, and Tartrazine in gummy bears, chewing gum, and sweet candy samples by reversed phase-high-performance liquid chromatography (RP-HPLC) coupled with UV detection
  • Construction of an electrochemical sensor based on cobalt complex decorated carbon paste electrode (CoC/CPE) for the cyclic voltametric determination of tartrazine, sunset yellow, and Allura red in gelatin and pharmaceutical dosages of acetaminophen
  • Multi-residue analysis of 20 synthetic dyes in 18 chili powders and six syrup-preserved fruit samples by liquid chromatography-tandem mass spectrometry (LC-MS/MS)
  • Spectrophotometric quantitative estimation of Brilliant Blue, Sunset Yellow, Ponceau 4R, and Tartrazin from juice samples, using a transparent polymer matrix for extraction
  • Liquid-liquid microextraction of five synthetic food dyes from different food samples for their determination by high-performance liquid chromatography with a variable wavelength ultraviolet detector (HPLC-VWUVD)

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mehrorang Ghaedi
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 94, 346-351 (2012-05-12)
Adsorption of Sunset yellow (SY) onto cadmium hydroxide nanowires loaded on activated carbon (Cd(OH)(2)-NW-AC) and silver nanoparticles loaded on activated carbon (Ag-NP-AC) was investigated. The effects of pH, contact time, amount of adsorbents, initial dye concentration, agitation speed and temperature
M Ghaedi et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 90, 22-27 (2012-02-07)
Adsorption is a promising technique for decolorization of effluents of textile dyeing industries but its application is limited due to requirement of high amounts of adsorbent required. The objective of this study was to assess the potential of cadmium telluride
Florin Soponar et al.
Journal of chromatography. A, 1188(2), 295-300 (2008-03-21)
A high-performance thin-layer chromatographic method combined with image processing of scanned chromatograms was developed for the determination of some food dyes (tartrazine, azorubine and Sunset Yellow) in different products. Porous silica gel with 3-aminopropyl functional groups attached to the matrix
Sheng Tian Huang et al.
The Analyst, 137(11), 2593-2599 (2012-05-01)
We report on a fast, sensitive, label-free, and general dye-sensor platform for synthetic organic dyes detection by competitive adsorption on reduced graphene oxide (rGO) against a fluorescent dye (FD). Fluorescein (Fl) as fluorescence indicator and a cationic dye methylene blue
Electro-Fenton oxidation of Sunset Yellow FCF azo-dye in aqueous solutions.
Ghoneim, Mohamed M., Hanaa S. El-Desoky, and Naglaa M. Zidan
Desalination, 274 (1), 22-30 (2011)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service