Skip to Content
Merck
All Photos(2)

Key Documents

64188

Supelco

D-Gluconic acid sodium salt

certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Synonym(s):

2,3,4,5,6-Pentahydroxycaproic acid sodium salt, D-Gluconate sodium salt, Sodium D-gluconate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H11NaO7
CAS Number:
Molecular Weight:
218.14
Beilstein:
3919651
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material
TraceCERT®

Quality Level

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

SMILES string

[Na+].OC[C@@H](O)[C@@H](O)[C@H](O)[C@@H](O)C([O-])=O

InChI

1S/C6H12O7.Na/c7-1-2(8)3(9)4(10)5(11)6(12)13;/h2-5,7-11H,1H2,(H,12,13);/q;+1/p-1/t2-,3-,4+,5-;/m1./s1

InChI key

UPMFZISCCZSDND-JJKGCWMISA-M

Looking for similar products? Visit Product Comparison Guide

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.
Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ubaid H Shah et al.
Clinical nutrition (Edinburgh, Scotland), 32(2), 193-199 (2012-09-18)
Acute lower respiratory infections are the most frequent illnesses globally in children less than 5 years old. The aim of this randomized double blind controlled trial is to assess the effectiveness of zinc gluconate supplementation for 2 months period compared
Yanshun Xu et al.
Food chemistry, 134(2), 1005-1010 (2012-10-31)
The structural changes and aggregation properties of silver carp actomyosin acidified with d-gluconic acid-δ-lactone (GDL) were investigated. Results showed that silver carp actomyosin underwent aggregation and formation of precipitate as indicated by turbidity and centrifugation coupled electrophoresis analysis. Circular dichroism
T Shigeta et al.
Transplantation proceedings, 44(4), 942-945 (2012-05-09)
The aims of this study were to compare extracellular and intracellular-type University of Wisconsin (UW) solutions for liver grafts and to assess oxygenation in this perfusion system. The organ preservation system consisted of 3 circulating systems for the portal vein
Volker Behrends et al.
The Journal of biological chemistry, 288(21), 15098-15109 (2013-04-11)
Metabolic footprinting of supernatants has been proposed as a tool for assigning gene function. We used NMR spectroscopy to measure the exometabolome of 86 single-gene transposon insertion mutant strains (mutants from central carbon metabolism and regulatory mutants) of the opportunistic
Hossam E Emam et al.
Carbohydrate polymers, 90(3), 1345-1352 (2012-09-04)
Copper containing cellulose material is of growing interest, e.g. offering alternative in the field of antimicrobials. Solutions of copper d-gluconate complexes (Cu(2+)-DGL) were used to introduce copper ions into a swollen cellulosic matrix. A ligand exchange mechanism forms the chemical

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service