Skip to Content
Merck
All Photos(1)

Key Documents

63089

Sigma-Aldrich

Magnesium oxide

BioUltra, ≥97.0% (calcined substance, KT)

Synonym(s):

Magnesia, MgO

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
MgO
CAS Number:
Molecular Weight:
40.30
EC Number:
MDL number:
UNSPSC Code:
12352303
PubChem Substance ID:
NACRES:
NA.25
Assay:
≥97.0% (calcined substance, KT)
solubility:
5 M HCl: 0.1 M at 20 °C, clear, colorless

product line

BioUltra

Quality Level

Assay

≥97.0% (calcined substance, KT)

impurities

insoluble matter, passes filter test
≤0.002% total nitrogen (N)

loss

≤3% loss on ignition, 900 °C

mp

2852 °C (lit.)

solubility

5 M HCl: 0.1 M at 20 °C, clear, colorless

anion traces

chloride (Cl-): ≤100 mg/kg
sulfate (SO42-): ≤10 mg/kg

cation traces

Al: ≤20 mg/kg
As: ≤0.1 mg/kg
Ba: ≤10 mg/kg
Bi: ≤10 mg/kg
Ca: ≤500 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤50 mg/kg
K: ≤50 mg/kg
Li: ≤50 mg/kg
Mn: ≤5 mg/kg
Mo: ≤5 mg/kg
Na: ≤2000 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Sr: ≤20 mg/kg
Zn: ≤5 mg/kg

λ

0.1 M in 5 M HCl

UV absorption

λ: 260 nm Amax: ≤0.040
λ: 280 nm Amax: ≤0.025

SMILES string

O=[Mg]

InChI

1S/Mg.O

InChI key

CPLXHLVBOLITMK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Magnesium oxide is used as a dietary supplement to study its effects on processes such as the rate of calcium uptate and ATPase activity. MgO is used in the development of various nanoparticles.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zi-Yu Wang et al.
International journal of nanomedicine, 6, 871-875 (2011-07-02)
To explore the preparation and characterization of a novel nanosized magnetic liposome containing the PEI-As(2)O(3)/Mn(0.5)Zn(0.5)Fe(2)O(4) complex. Mn(0.5)Zn(0.5)Fe(2)O(4) and As(2)O(3)/Mn(0.5)Zn(0.5)Fe(2)O(4) nanoparticles were prepared by chemical coprecipitation and loaded with PEI. The PEI- As(2)O(3)/Mn(0.5)Zn(0.5)Fe(2)O(4) complex was characterized using transmission electron and scanning
Verena Wilhelmi et al.
Toxicology in vitro : an international journal published in association with BIBRA, 26(2), 323-334 (2011-12-27)
Current hazard characterisation of nanoparticles (NP) is predominantly based on in vitro test systems, being established for small molecules of drugs and chemicals. However, specific physicochemical properties of NP may result in interference with assay components, biomarkers, or detection systems.
Benjamin Michen et al.
Environmental science & technology, 47(3), 1526-1533 (2013-01-05)
Ceramic filters, working on the depth filtration principle, are known to improve drinking water quality by removing human pathogenic microorganisms from contaminated water. However, these microfilters show no sufficient barrier for viruses having diameters down to 20 nm. Recently, it
Charles T Campbell et al.
Journal of the American Chemical Society, 134(43), 18109-18115 (2012-10-05)
Adsorbed molecules are involved in many reactions on solid surface that are of great technological importance. As such, there has been tremendous effort worldwide to learn how to predict reaction rates and equilibrium constants for reactions involving adsorbed molecules. Theoretical
C B Abi et al.
Journal of the mechanical behavior of biomedical materials, 18, 123-131 (2012-12-25)
The aim of the present study was to investigate the production of tetragonal zirconia (t-ZrO(2)) particles (experimental t-ZrO(2)) from monoclinic zirconia (m-ZrO(2)) and to evaluate the effect of the t-ZrO(2) content on the fracture toughness of alumina-zirconia composites by conducting

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service