Skip to Content
Merck

Skip To

61849

2-[4-(2-Methylpropyl)phenyl]ethanol

pharmaceutical impurity standard

Synonym(s):

Ibuprofen impurity Q (PhEur), 4-(2-Methylpropyl)benzeneethanol, 4-Isobutylphenylethanol, NSC 69112

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C12H18O
CAS Number:
Molecular Weight:
178.27
UNSPSC Code:
41116107
NACRES:
NA.24
PubChem Substance ID:
MDL number:
Beilstein/REAXYS Number:
1866730
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

API family

ibuprofen

assay

≥98.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

impurities

≤1% water

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

OCCC1=CC=C(CC(C)C)C=C1

InChI

1S/C12H18O/c1-10(2)9-12-5-3-11(4-6-12)7-8-13/h3-6,10,13H,7-9H2,1-2H3

InChI key

CBKUDUSUCXBKIE-UHFFFAOYSA-N



Still not finding the right product?


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters