Skip to Content
Merck

61706

Linalool

certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Synonym(s):

(±)-3,7-Dimethyl-1,6-octadien-3-ol, (±)-3,7-Dimethyl-3-hydroxy-1,6-octadiene

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
(CH3)2C=CHCH2CH2C(CH3)(OH)CH=CH2
CAS Number:
Molecular Weight:
154.25
UNSPSC Code:
41116107
NACRES:
NA.24
PubChem Substance ID:
EC Number:
201-134-4
Beilstein/REAXYS Number:
1721488
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

vapor pressure

0.17 mmHg ( 25 °C)

grade

certified reference material, TraceCERT®

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

refractive index

n20/D 1.462 (lit.)

bp

194-197 °C/720 mmHg (lit.)

density

0.87 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
environmental
flavors and fragrances
food and beverages
personal care

format

neat

storage temp.

−20°C

SMILES string

C\C(C)=C\CCC(C)(O)C=C

InChI

1S/C10H18O/c1-5-10(4,11)8-6-7-9(2)3/h5,7,11H,1,6,8H2,2-4H3

InChI key

CDOSHBSSFJOMGT-UHFFFAOYSA-N

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.
Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com.

Other Notes

This compound is commonly found in plants of the genus: cinnamomum salvia thymus

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany


Still not finding the right product?

Explore all of our products under Linalool


pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1

Storage Class

10 - Combustible liquids

wgk

WGK 1

flash_point_f

171.0 °F - Pensky-Martens closed cup

flash_point_c

77.2 °C - Pensky-Martens closed cup



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library