Skip to Content
Merck

61328

Lactoperoxidase from bovine milk

lyophilized, powder, ≥150 U/mg

Synonym(s):

Donor:hydrogen peroxide oxidoreductase, Peroxidase

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

CAS Number:
NACRES:
NA.22
UNSPSC Code:
12352204
EC Number:
232-668-6
MDL number:
EC Number:
Specific activity:
≥150 U/mg
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

powder

Quality Segment

quality

lyophilized

specific activity

≥150 U/mg

mol wt

Mr ~77500

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

SMILES string

[O+H2]O[O-]

InChI

1S/H2O3/c1-3-2/h1-2H

InChI key

JSPLKZUTYZBBKA-UHFFFAOYSA-N

Gene Information

cow ... LPO(280844)

Biochem/physiol Actions

Lactoperoxidase catalyzes the oxidation of iodide to iodine by hydrogen peroxide. This activity provides a gentle, specific alternative to chloramine T for the radioiodination of proteins and DNA.

Other Notes

1 U corresponds to the amount of enzyme which oxidizes 1 μmol ABTS (Cat. No. 11557) per minute at pH 6.0 and 25°C
Sales restrictions may apply
Lactoperoxidase system of bovine milk, review; Structural and functional properties of; Products of lactoperoxidase-catalyzed oxidation of thiocyanate and halides


Still not finding the right product?

Explore all of our products under Lactoperoxidase from bovine milk


pictograms

Health hazard

signalword

Danger

hcodes

Hazard Classifications

Resp. Sens. 1

Storage Class

11 - Combustible Solids

wgk

WGK 1

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library







Global Trade Item Number

SKUGTIN
61328-1MG-F04061832699714