Skip to Content
Merck

60744

Silica gel Inorganic Sorbent

high-purity grade, 550-950 mesh

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

CAS Number:
UNSPSC Code:
23201100
PubChem Substance ID:
NACRES:
SB.52
EC Number:
231-545-4
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Silica gel, high-purity grade, suitable for column chromatography, 90Å, 15-25 μm

grade

high-purity grade

Quality Segment

form

powder

technique(s)

LPLC: suitable

surface area

400 m2/g , B.E.T

matrix

Silica

matrix active group

silica

particle size

0.015-0.025 mm, 15-25 μm

pore size

90 Å mean pore size

bp

2230 °C

mp

>1600 °C

suitability

suitable for column chromatography

separation technique

hydrophilic interaction (HILIC)

SMILES string

O=[Si]=O

InChI

1S/O2Si/c1-3-2

InChI key

VYPSYNLAJGMNEJ-UHFFFAOYSA-N

General description

Silica gel is a rigid 3D network of colloidal silica. It is exclusively used as a support material for the active titanium (III) centers in Ziegler-Natta catalysts. Silica gel allows a controlled fragmentation leading to form uniform polymer particles with narrow particle size distribution and high bulk density. Silica gel is classified as below:
a. Aquagel - pores are filled with water
b. Xerogel - by the process of evaporation, aqueous phase in the pores are removed
c. Aerogel - Solvent removed by supercritical extraction.

Application

Silica gel may be used in separation of single-walled carbon nanotube (SWNT) by simple filtration.


Still not finding the right product?

Explore all of our products under Silica gel


Storage Class

11 - Combustible Solids

wgk

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library