60386
Potassium iodate
reference material for titrimetry, certified by BAM, >99.5%
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
grade
reference material
Quality Level
Assay
>99.5%
quality
certified by BAM
technique(s)
titration: suitable
mp
560 °C (lit.)
density
3.93 g/mL at 25 °C (lit.)
format
neat
SMILES string
[K+].[O-]I(=O)=O
InChI
1S/HIO3.K/c2-1(3)4;/h(H,2,3,4);/q;+1/p-1
InChI key
JLKDVMWYMMLWTI-UHFFFAOYSA-M
Looking for similar products? Visit Product Comparison Guide
General description
Potassium iodate is a volumetric standard manufactured under stringent conditions and measured with the highest possible precision. Its content determination and homogeneity assessment are performed by volumetric redox titration as the primary analysis method.
Application
It is used as an oxidimetric standard for the standardization of volumetric solutions in iodometric titrations.
Features and Benefits
- Available as a solid in a secure glass bottle to ensure its stability for the entire shelf life until opened.
- High-purity material traceable to NIST SRM
- High-quality standard offering accurate titer determinations
- Accompanied by a certificate of analysis (CoA)
Analysis Note
Exact concentration, expiry date and detailed information on certification can be found on the certificate and certification report, included in each package. Content and expiry date on label.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Eye Irrit. 2 - Ox. Sol. 2
Storage Class Code
5.1B - Oxidizing hazardous materials
WGK
WGK 1
Flash Point(F)
does not flash
Flash Point(C)
does not flash
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Talanta, 78(2), 584-589 (2009-02-11)
In this work, an on-line system with vapor-phase generation (VPG) and Fourier transform infrared (FTIR) spectrometric detection has been developed as a direct and highly selective analytical technique for the assay of penicillamine (PA). Potassium iodate solution was injected into
Academic radiology, 18(6), 682-689 (2011-03-12)
Evaluation of contrast enhancement of pulmonary lesions with ground-glass attenuation (GGA) is difficult with conventional techniques but might be possible using contrast-mapping images (CMIs) obtained by dual-energy computed tomography. To address this issue, a phantom study was conducted, and this
Journal of perinatology : official journal of the California Perinatal Association, 27(5), 320-322 (2007-04-25)
Congenital thyrotoxicosis is a rare and potentially fatal illness. We report a case in a preterm infant delivered to a mother known to have autoimmune endocrine disease. Diagnosis was difficult because the infant's presenting symptoms and signs closely resembled congenital
Journal of trace elements in medicine and biology : organ of the Society for Minerals and Trace Elements (GMS), 22(1), 1-8 (2008-03-06)
Iodine deficiency disorders continue to be a severe problem in many parts of Central Asia, causing delayed mental development and cretinism in indigenous populations. In some areas, iodized salt has not succeeded in controlling this problem. In southern Xinjiang Province
Nutrition journal, 9, 68-68 (2010-12-21)
Evidence showed that the occurrence of iodine deficiency endemic areas has been found in every provinces of Thailand. Thus, a new pilot programme for elimination of iodine deficiency endemic areas at the community level was designed in 2008 by integrating
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service