Skip to Content
Merck
All Photos(1)

Documents

60372

Sigma-Aldrich

Secoisolariciresinol

≥95.0% (HPLC)

Synonym(s):

(2R,3R)-2,3-Bis(4-hydroxy-3-methoxybenzyl)-1,4-butanediol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H26O6
Molecular Weight:
362.42
Beilstein:
6611290
EC Number:
MDL number:
UNSPSC Code:
41116105
PubChem Substance ID:
NACRES:
NA.21

Quality Level

Assay

≥95.0% (HPLC)

form

solid

SMILES string

OC[C@@H]([C@H](CO)CC1=CC=C(O)C(OC)=C1)CC2=CC(OC)=C(O)C=C2

InChI

1S/C20H26O6/c1-25-19-9-13(3-5-17(19)23)7-15(11-21)16(12-22)8-14-4-6-18(24)20(10-14)26-2/h3-6,9-10,15-16,21-24H,7-8,11-12H2,1-2H3/t15-,16-/m0/s1

InChI key

PUETUDUXMCLALY-HOTGVXAUSA-N

Application

Secoisolariciresinol is a metabolite of secoisolariciresinol diglucoside (SDG), an antioxidant present in flaxseed.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Physical form

mixture of enantiomers of the (R*,R*)-diastereoisomer

Other Notes

Component of functional food. Flaxseed lignan with antioxidant activity

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hisashi Nishiwaki et al.
Bioscience, biotechnology, and biochemistry, 75(9), 1735-1739 (2011-09-08)
The larvicidal activity against Culex pipiens of all stereoisomers of dihydroguaiaretic acid (DGA) and secoisolariciresinol was measured, and these DGAs were found to be potent. Sixteen (-)-DGA derivatives were then newly synthesized to analyze their structure-activity relationship. Two derivatives monohydroxylated
Carol J Fabian et al.
Cancer prevention research (Philadelphia, Pa.), 3(10), 1342-1350 (2010-08-21)
Preclinical and correlative studies suggest reduced breast cancer with higher lignan intake or blood levels. We conducted a pilot study of modulation of risk biomarkers for breast cancer in premenopausal women after administration of the plant lignan secoisolariciresinol given as
M R O'Neil et al.
Domestic animal endocrinology, 37(3), 148-158 (2009-06-30)
To evaluate the estrogenic potential of secoisolariciresinol diglycoside (SDG) found in linseed meal (LSM) on visceral organ mass, IGF-I, and thyroid hormone (T(3) and T(4)) concentrations, 48 multiparous, ovariectomized ewes (54.6 +/- 1.1 kg) were used in a 3 x
Niina M Saarinen et al.
The British journal of nutrition, 104(6), 833-841 (2010-04-15)
Limited information is available on lignan metabolism and tissue distribution between sexes and the effects of prolonged lignan exposure on tissue concentrations. In the present study, excretion and tissue distribution of lignans were compared after 1 d and 7 d
K Struijs et al.
Journal of applied microbiology, 107(1), 308-317 (2009-03-24)
It has been investigated whether secoisolariciresinol (SECO) and anhydrosecoisolariciresinol (AHS), an acid degradation product of SECO, could be fermented in a similar way, and to a similar extent, by members of the intestinal microbiota. AHS and SECO were demethylated by

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service