Skip to Content
Merck
All Photos(1)

Key Documents

55370

Supelco

8-Hydroxy-7-iodo-5-quinolinesulfonic acid

for spectrophotometric det. of Fe(III), ≥98.5%

Synonym(s):

Ferron, Iodoxyquinolinesulfonic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H6INO4S
CAS Number:
Molecular Weight:
351.12
Beilstein:
223832
EC Number:
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:
NACRES:
NA.21

Quality Level

Assay

≥98.5% (T)
≥98.5%

quality

for spectrophotometric det. of Fe(III)

technique(s)

UV/Vis spectroscopy: suitable

mp

269-270 °C (dec.) (lit.)

SMILES string

Oc1c(I)cc(c2cccnc12)S(O)(=O)=O

InChI

1S/C9H6INO4S/c10-6-4-7(16(13,14)15)5-2-1-3-11-8(5)9(6)12/h1-4,12H,(H,13,14,15)

InChI key

ZBJWWKFMHOAPNS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Reagent for the extraction and spectrophotometric determination of Fe

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D Wang et al.
Water research, 35(14), 3418-3428 (2001-09-08)
As a new kind of water and wastewater treatment reagents, inorganic polymer flocculants (IPFs) are in a trend of rapid development. Among them, iron-based IPF without any toxic problems is greatly expected. In this paper, a new protocol for preparation
G. Przeszlakowski et al.
Analyst, 107, 1320-1320 (1982)
Role of ferric iron in platelet lipoxygenase activity.
J E Greenwald et al.
Biochemical and biophysical research communications, 96(2), 817-822 (1980-09-30)
M Saeed et al.
European radiology, 8(6), 1047-1053 (1998-07-31)
Our objective was: (1) to determine the appropriate dose of new ultrasmall superparamagnetic iron oxide particles for magnetic resonance angiography (MRA). This agent comprised of a single iron oxide crystal stabilized with a carbohydrate-polyethylene glycol coat (PEG-Ferron/NC 100150 injection); (2)
Ye Changqing et al.
Water science and technology : a journal of the International Association on Water Pollution Research, 59(4), 823-832 (2009-02-25)
The speciation of Al-OH complexes in terms of Al(a), Al(b) and Al(c) could be achieved by traditional ferron assay and Al(b) is generally considered as Al(13), however, the inherent correlation between them remains an enigma. This paper presents a modified

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service