Skip to Content
Merck
All Photos(2)

Key Documents

53889

Supelco

Bacoside A3

analytical standard

Synonym(s):

(3β,16β,23R)-16,23:16,30-Diepoxy-20-hydroxydammar-24-en-3-yl O-α-L-arabinofuranosyl-(1→2)-O-[β-D-glucopyranosyl-(1→3)]-β-D-glucopyranoside, Jujubogenin 3-O-[α-L-arabinofuranosyl-(1→2)-O-[β-D-glucopyranosyl-(1→3)]-β-D-glucopyranoside]

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C47H76O18
CAS Number:
Molecular Weight:
929.10
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥95% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

food and beverages

format

neat

SMILES string

C\C(C)=C\[C@H]1C[C@](C)(O)[C@@H]2[C@H]3CC[C@@H]4[C@@]5(C)CC[C@H](O[C@@H]6O[C@H](CO)[C@@H](O)[C@H](O[C@@H]7O[C@H](CO)[C@@H](O)[C@H](O)[C@H]7O)[C@H]6O[C@@H]8O[C@@H](CO)[C@H](O)[C@H]8O)C(C)(C)[C@@H]5CC[C@@]4(C)[C@@]39CO[C@@]2(C9)O1

InChI

1S/C47H76O18/c1-21(2)14-22-15-45(7,57)38-23-8-9-28-43(5)12-11-29(42(3,4)27(43)10-13-44(28,6)46(23)19-47(38,65-22)58-20-46)62-41-37(64-39-34(55)31(52)25(17-49)60-39)36(32(53)26(18-50)61-41)63-40-35(56)33(54)30(51)24(16-48)59-40/h14,22-41,48-57H,8-13,15-20H2,1-7H3/t22-,23+,24+,25-,26+,27-,28+,29-,30+,31-,32+,33-,34+,35+,36-,37+,38-,39-,40-,41-,43-,44+,45-,46-,47-/m0/s1

InChI key

CDEVGTJBRPBOPH-INTDMYAHSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Bacoside A3 is a triterpenoid saponin and an important constituent of bacosides, a saponin mixture of Bacopa monniera.

Application

Bacoside A3 may be used as a reference standard in the determination of bacoside A3 in Bacopa monnieri (L.) Wettst. extract using reversed-phase high performance liquid chromatography (RP-HPLC).
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sukanya Majumdar et al.
Plant cell reports, 31(10), 1899-1909 (2012-06-27)
Genetic transformation of the Indian medicinal plant, Bacopa monnieri, using a gene encoding cryptogein, a proteinaceous elicitor, via Ri and Ti plasmids, were established and induced bioproduction of bacopa saponins in crypt-transgenic plants were obtained. Transformed roots obtained with A.
R Pawar et al.
Planta medica, 67(8), 752-754 (2001-12-04)
The hydroalcoholic extract of the whole plant of Bacopa monniera Wettst. (Scrophulariaceae), exhibited an inhibitory effect on superoxide released from polymorphonuclear (PMN) cells in the nitroblue tetrazolium (NBT) assay. The major saponin bacoside A(3) was found to be responsible for
K Sairam et al.
Phytomedicine : international journal of phytotherapy and phytopharmacology, 8(6), 423-430 (2002-02-05)
Bacopa monniera Wettst. (BM, syn. Herpestis monniera L; Scrophulariaceae), is an Ayurvedic drug used as a rasayana. Its fresh juice was earlier reported to have significant antiulcerogenic activity. In continuation, methanolic extract of BM (BME) standardized to bacoside-A content (percentage-38.0
Charinrat Tothiam et al.
Phytochemical analysis : PCA, 22(5), 385-391 (2011-03-18)
In Ayurvedic medicines, Bacopa monnieri (L.) Wettst. (brahmi) is known as a medicinal plant used for memory enhancement. Its active compounds are classified as pseudojujubogenin and jujubogenin glycosides. Owing to the lack of chromophore in the saponin glycoside structures, HPLC-UV-vis
Khalid Rauf et al.
Phytomedicine : international journal of phytotherapy and phytopharmacology, 18(10), 836-842 (2011-03-08)
Opioids are extensively used for the management of both chronic malignant and non malignant pains. One major serious limitation associated with chronic use of opioids is the development of tolerance to its analgesic effect. The effect of Bacopa monnieri, a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service