Skip to Content
Merck
All Photos(1)

Key Documents

51411

Supelco

Fluorobenzene

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H5F
CAS Number:
Molecular Weight:
96.10
Beilstein:
1236623
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥99.7% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.465 (lit.)
n20/D 1.465

bp

85 °C (lit.)

mp

−42 °C (lit.)

density

1.024 g/mL at 25 °C (lit.)

application(s)

environmental

format

neat

SMILES string

Fc1ccccc1

InChI

1S/C6H5F/c7-6-4-2-1-3-5-6/h1-5H

InChI key

PYLWMHQQBFSUBP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Fluorobenzene is a colorless liquid, which can find applications as a reagent for resin or plastic polymers and also as an insecticide. It also plays the role of a capping agent in the synthesis of a non-fullerene electron acceptor like SF(DPPFB)4 comprising of a spirobifluorene core and four diketopyrrolopyrrole (DPP) arms that can find applications in designing organic solar cells.

Application

Fluorobenzene may be used as an analytical reference standard for the determination of the analyte in cell suspensions of Rhizobiales strain F11, and water samples by liquid chromatography (LC) based techniques. It may also be used as an internal standard for the determination of volatile aromatic compounds in gasoline by comprehensive two-dimensional gas chromatography (GC x GC).
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Eye Dam. 1 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

5.0 °F - closed cup

Flash Point(C)

-15 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A non-fullerene electron acceptor with a spirobifluorene core and four diketopyrrolopyrrole arms end capped by 4-fluorobenzene
Yuan C, et al.
Dyes and Pigments, 143, 217-222 (2017)
Maria F Carvalho et al.
Applied and environmental microbiology, 72(11), 7413-7417 (2006-09-19)
The aerobic metabolism of fluorobenzene by Rhizobiales sp. strain F11 was investigated. Liquid chromatography-mass spectrometry analysis showed that 4-fluorocatechol and catechol were formed as intermediates during fluorobenzene degradation by cell suspensions. Both these compounds, unlike 3-fluorocatechol, supported growth and oxygen
Sittig's Handbook of Toxic and Hazardous Chemicals and Carcinogens, Volume 1 (2017)
Quantitative determination of BTEX and total aromatic compounds in gasoline by comprehensive two-dimensional gas chromatography (GC x GC)
Frysinger GS, et al.
Journal of High Resolution Chromatography, 22(4), 195-200 (1999)
A Yokota-Nakatsuma et al.
Mucosal immunology, 7(4), 786-801 (2013-11-14)
The vitamin A (VA) metabolite retinoic acid (RA) affects the properties of T cells and dendritic cells (DCs). In VA-deficient mice, we observed that mesenteric lymph node (MLN)-DCs induce a distinct inflammatory T helper type 2 (Th2)-cell subset that particularly

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service