Skip to Content
Merck
All Photos(2)

Key Documents

50843

Supelco

D-Camphor

analytical standard

Synonym(s):

2-Bornanone, 2-Camphanone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H16O
CAS Number:
Molecular Weight:
152.23
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

Quality Level

grade

analytical standard

vapor density

5.24 (vs air)

vapor pressure

4 mmHg ( 70 °C)

Assay

≥97.5% (GC)

optical activity

[α]/D +43.0±2.0°, c = 10 in ethanol

autoignition temp.

870 °F

shelf life

limited shelf life, expiry date on the label

expl. lim.

3.5 %

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

178-182 °C (lit.)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

SMILES string

CC1(C)[C@@H]2CC[C@@]1(C)C(=O)C2

InChI

1S/C10H16O/c1-9(2)7-4-5-10(9,3)8(11)6-7/h7H,4-6H2,1-3H3/t7-,10+/m1/s1

InChI key

DSSYKIVIOFKYAU-XCBNKYQSSA-N

Looking for similar products? Visit Product Comparison Guide

General description

D-Camphor is an acyclic terpenoid, that occurs naturally in the wood of Cinnamomum camphora.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Eye Dam. 1 - Flam. Sol. 2 - Skin Irrit. 2 - STOT SE 2 Inhalation

Target Organs

Lungs

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 1

Flash Point(F)

147.2 °F - closed cup

Flash Point(C)

64 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The Pharmacology of Chinese Herbs, Second Edition (1998)
R E White et al.
Archives of biochemistry and biophysics, 228(2), 493-502 (1984-02-01)
Three alicyclic compounds (D-camphor, adamantanone, adamantane) were found to be hydroxylated by the cytochrome P-450 isoenzymes P-450cam and P-450LM2. With P-450cam as the catalyst only one product was formed from each of the substrates: 5-exohydroxycamphor, 5-hydroxyadamantanone, and 1-adamantanol. With P-450LM2
Jeffrey N Love et al.
The Journal of emergency medicine, 27(1), 49-54 (2004-06-29)
Serious pediatric toxicity resulting from exposure to small amounts of camphor-containing products has long been a problem. Twenty years ago the United States Food and Drug Administration took several actions in an attempt to ameliorate this risk. Despite these changes
J Vilaplana et al.
Actas dermo-sifiliograficas, 98(5), 345-346 (2007-06-09)
We report a case of allergic contact dermatitis due to alcamphor present in a flushing solution used by a 58-year-old woman. It is the first case described in our Division of Allergy and the second one reported in the literature.
E Siegel et al.
Pediatric clinics of North America, 33(2), 375-379 (1986-04-01)
Camphor is present in several over-the-counter compounds of questionable use and therefore may be ingested by small children. Because seizures may follow ingestion of certain amounts, appropriate treatment is needed, including the use of anticonvulsants.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service