Skip to Content
Merck

Skip To

46702

Pyrantel pamoate

VETRANAL®, analytical standard

Synonym(s):

1-Methyl-2-(2-[2-thienyl]ethenyl)-1,4,5,6-tetrahydropyrimidine compound with 4,4′-methylenebis(3-hydroxy-2-naphthoic acid), Pyrantel embonate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Linear Formula:
C11H14N2S · C23H16O6
CAS Number:
Molecular Weight:
594.68
UNSPSC Code:
41116107
NACRES:
NA.24
PubChem Substance ID:
EC Number:
244-837-1
Beilstein/REAXYS Number:
9034755
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

analytical standard

Quality Segment

product line

VETRANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

application(s)

clinical testing

format

neat

SMILES string

CN1CCCN=C1\C=C\c2cccs2.OC(=O)c3cc4ccccc4c(Cc5c(O)c(cc6ccccc56)C(O)=O)c3O

InChI

1S/C23H16O6.C11H14N2S/c24-20-16(14-7-3-1-5-12(14)9-18(20)22(26)27)11-17-15-8-4-2-6-13(15)10-19(21(17)25)23(28)29;1-13-8-3-7-12-11(13)6-5-10-4-2-9-14-10/h1-10,24-25H,11H2,(H,26,27)(H,28,29);2,4-6,9H,3,7-8H2,1H3/b;6-5+

InChI key

AQXXZDYPVDOQEE-MXDQRGINSA-N

General description

Chemical structure: tetrahydropyrimidine

Application

Pyrantel pamoate may be used as a reference standard in the determination of pyrantel pamoate in dog plasma and veterinary tablets using high performance liquid chromatography (HPLC) and mid-infrared spectroscopy (IR), respectively.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
1584003PHR2080Y0000402
technique(s)

HPLC: suitable, gas chromatography (GC): suitable

technique(s)

-

technique(s)

-

technique(s)

-

format

neat

format

neat

format

neat

format

neat

Quality Level

200

Quality Level

-

Quality Level

300

Quality Level

-

grade

analytical standard

grade

pharmaceutical primary standard

grade

certified reference material, pharmaceutical secondary standard

grade

pharmaceutical primary standard

shelf life

limited shelf life, expiry date on the label

shelf life

-

shelf life

-

shelf life

-

product line

VETRANAL®

product line

-

product line

-

product line

-


Still not finding the right product?

Explore all of our products under Pyrantel pamoate


Storage Class

11 - Combustible Solids

wgk

WGK 3

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters