Skip to Content
Merck
All Photos(1)

Key Documents

46036

Supelco

Brodifacoum

PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C31H23BrO3
CAS Number:
Molecular Weight:
523.42
Beilstein:
8171403
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

OC1=C(C2CC(Cc3ccccc23)c4ccc(cc4)-c5ccc(Br)cc5)C(=O)Oc6ccccc16

InChI

1S/C31H23BrO3/c32-24-15-13-20(14-16-24)19-9-11-21(12-10-19)23-17-22-5-1-2-6-25(22)27(18-23)29-30(33)26-7-3-4-8-28(26)35-31(29)34/h1-16,23,27,33H,17-18H2

InChI key

VEUZZDOCACZPRY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Brodifacoum is an anticoagulant rodenticide, widely used for the control of rodent population via inhibition of the vitamin K epoxide reductase, causing blood clotting alterations, leading to haemorrhaging as the ultimate cause of death.[1]

Application

Brodifacoum may be used as an analytical reference standard for the quantification of the analyte in animal samples[1] and biological samples[2] using different chromatography techniques..
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 1 Inhalation - Acute Tox. 1 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Repr. 1A - STOT RE 1

Target Organs

Blood

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

The determination of the anticoagulant rodenticide brodifacoum in blood serum by liquid chromatography with fluorescence detection
Felice.JL and Murphy.JM
Journal of Analytical Toxicology, 13(4), 229-231 (1989)
Reversed-phase HPLC determination of eight anticoagulant rodenticides in animal liver
Fauconnet.V, et al.
Journal of Analytical Toxicology, 21(7), 548-553 (1997)
Naren Gunja et al.
Clinical toxicology (Philadelphia, Pa.), 49(5), 385-390 (2011-07-12)
Brodifacoum is a widely available superwarfarin used as a commercial rodenticide. Toxicity from long-acting anticoagulant rodenticides, primarily from uncontrolled bleeding, has been reported. Very little published toxicokinetic data are available for human brodifacoum poisoning. Management is also contentious with uncertainty
Manifestations of superwarfarin ingestion: a plea to increase awareness.
Dipika Misra et al.
American journal of hematology, 85(5), 391-392 (2010-03-17)
Kaylin S Watson et al.
The Journal of the Louisiana State Medical Society : official organ of the Louisiana State Medical Society, 164(2), 70-72 (2012-06-13)
Superwarfarin vitamin K antagonists are found in rat poisons and are readily available. Pediatric exposures are common but are usually asymptomatic without significant coagulopathy. Superwarfarin intoxication must be considered in any adult who presents with an unexplained coagulopathy with extreme

Questions

1–4 of 4 Questions  
  1. What is the purity of Product 46036, Brodifacoum?

    1 answer
    1. This product does not have a purity specification. However, the purity is provided on the certificate of analysis of each lot.

      Helpful?

  2. What is the Department of Transportation shipping information for this product?

    1 answer
    1. Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.

      Helpful?

  3. For what applications can Product 46036, Brodifacoum, be used?

    1 answer
    1. This product consists of a coumarin analog. It has anticoagulant activity and has been described as a rodenticide.

      Helpful?

  4. What is Product 46036, Brodifacoum, soluble in?

    1 answer
    1. This product is insoluble in water. It is slightly soluble in alcohol and benzene. It is soluble in acetone and chloroform.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service