Skip to Content
Merck
All Photos(1)

Documents

45802

Supelco

p-Terphenyl

analytical standard

Synonym(s):

1,4-Diphenylbenzene, PT, PTP

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5C6H4C6H5
CAS Number:
Molecular Weight:
230.30
Beilstein:
1908447
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

vapor density

7.95 (vs air)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

bp

389 °C (lit.)

mp

212-213 °C (lit.)

application(s)

environmental

format

neat

SMILES string

c1ccc(cc1)-c2ccc(cc2)-c3ccccc3

InChI

1S/C18H14/c1-3-7-15(8-4-1)17-11-13-18(14-12-17)16-9-5-2-6-10-16/h1-14H

InChI key

XJKSTNDFUHDPQJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 4 - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

404.6 °F - closed cup

Flash Point(C)

207 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Aurélia Caputo et al.
Biology direct, 10, 5-5 (2015-04-19)
Alterations in gut microbiota composition under antibiotic pressure have been widely studied, revealing a restricted diversity of gut flora, including colonization by organisms such as Enterococci, while their impact on bacterial load is variable. High-level colonization by Akkermansia muciniphila, ranging
Thomas A Lanz et al.
PloS one, 10(3), e0121744-e0121744 (2015-03-19)
Increased protein levels of striatal-enriched tyrosine phosphatase (STEP) have recently been reported in postmortem schizophrenic cortex. The present study sought to replicate this finding in a separate cohort of postmortem samples and to extend observations to striatum, including subjects with
Jonathan Breeze et al.
Nature communications, 6, 6215-6215 (2015-02-24)
Recently, the world's first room-temperature maser was demonstrated. The maser consisted of a sapphire ring housing a crystal of pentacene-doped p-terphenyl, pumped by a pulsed rhodamine-dye laser. Stimulated emission of microwaves was aided by the high quality factor and small
Franziska Mußbach et al.
Journal of cancer research and clinical oncology, 141(5), 813-825 (2014-11-07)
There is growing evidence for a role of proteinase-activated receptors (PARs), a subfamily of G protein-coupled receptors, in cancer. We have previously shown that PAR1 and PAR4 are able to promote the migration of hepatocellular carcinoma (HCC) cells suggesting a
Xiaojie Zhou et al.
PloS one, 9(9), e106932-e106932 (2014-09-30)
The German cockroach, Blattella germanica, is an important insect pest that transmits various pathogens mechanically and causes severe allergic diseases. This insect has long served as a model system for studies of insect biology, physiology and ecology. However, the lack

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service