Skip to Content
Merck

45554

Maneb

PESTANAL®, analytical standard

Synonym(s):

Manganese ethylenebis(dithiocarbamate)

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
[C4H6MnN2S4]n
CAS Number:
UNSPSC Code:
41116107
NACRES:
NA.24
PubChem Substance ID:
EC Number:
235-654-8
Beilstein/REAXYS Number:
3747585
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

analytical standard

Quality Segment

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

mol wt

Mw 265.30 (as monomer)

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

storage temp.

2-8°C

SMILES string

S=C1NCCNC(=S)S[Mn]S1

InChI

1S/C4H8N2S4.Mn/c7-3(8)5-1-2-6-4(9)10;/h1-2H2,(H2,5,7,8)(H2,6,9,10);/q;+2/p-2

InChI key

YKSNLCVSTHTHJA-UHFFFAOYSA-L

General description

Maneb is a manganese containing wide spectrum fungicide that belongs to the class of ethylene bis-dithiocarbamate (EBDC) compounds. It is commonly used to protect crops such as apples, pears, tobacco, grains, and beets.

Application

The maneb analytical standard can be used:

  • For the simultaneous determination of maneb along with its main metabolites from tomatoes using liquid chromatography
  • In the supramolecular liquid phase microextraction based UV-Vis spectrophotometric determination of maneb in food and water samples
  • For the spectrophotometric determination of maneb with 1-(2′-Pyridylazo)-2-naphthol (PAN)
  • To determine Maneb, Zineb, and their decomposition product with 2,6-dibromoquinone chlorimide (DBQ) and 2,6-dichloroquinone chlorimide (DCQ) by spectrophotometry

Other Notes

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany


signalword

Warning

Hazard Classifications

Acute Tox. 4 Inhalation - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Repr. 2 - Skin Sens. 1

Storage Class

4.2 - Pyrophoric and self-heating hazardous materials

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Faceshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Mustafa Soylak et al.
Food chemistry, 338, 128068-128068 (2020-09-20)
A novel and green method was developed for enrichment of maneb (manganese ethylene-bisdithiocarbamate) with a supramolecular solvent liquid phase microextraction method. The microextraction method has been used for the first time in the literature for separation-preconcentration of maneb. 1-decanol and
G Meco et al.
Scandinavian journal of work, environment & health, 20(4), 301-305 (1994-08-01)
Permanent parkinsonism was observed in a man with chronic exposure to the fungicide maneb (manganese ethylene-bis-dithiocarbamate). Symptoms developed at 37 years of age, two years after exposure had ceased. To our knowledge, this is the second report on parkinsonism associated
A L Rao et al.
Talanta, 40(2), 201-203 (1993-02-01)
A rapid, sensitive, simple and selective spectrophotometric method has been developed for the determination of micro-quantities of maneb (manganese ethylenebisdithiocarbamate) after extraction of the manganese-PAN complex in isobutyl methyl ketone (MIBK). The complex absorbs strongly at 550 nm. Beer's law



Global Trade Item Number

SKUGTIN
45554-250MG04061834044956