Skip to Content
Merck
All Photos(1)

Key Documents

45482

Supelco

Fenaminosulf

PESTANAL®, analytical standard

Synonym(s):

4-Dimethylaminobenzenediazosulfonic acid sodium salt, p-Diazodimethylaniline sodium sulfonate, Sodium [4-(dimethylamino)phenyl]diazenesulfonate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C8H10N3O3SNa
CAS Number:
Molecular Weight:
251.24
Beilstein:
4170376
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

[Na+].CN(C)c1ccc(cc1)N=NS([O-])(=O)=O

InChI

1S/C8H11N3O3S.Na/c1-11(2)8-5-3-7(4-6-8)9-10-15(12,13)14;/h3-6H,1-2H3,(H,12,13,14);/q;+1/p-1

InChI key

IWDQPCIQCXRBQP-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
Used in biological and pharmacological studies including:
  • Cloning and characterization of GABA transporter
  • Activity of anti-RSV carbonucleosides
  • Inhibition by fungicides of urea hydrolysis and nitrification of urea nitrogen in soil

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Aquatic Chronic 3

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P Stojanov et al.
Pacing and clinical electrophysiology : PACE, 21(1 Pt 1), 65-68 (1998-02-25)
Endocardial pacing system implantation has been performed in 15 children of mean age 37 months (ranging from 1 day to 89 months). Endocardial lead fixation was performed by means of slowly resorbable suture (Dexon) to allow spontaneous lead migration as
S Vainionpää et al.
Biomaterials, 8(1), 46-48 (1987-01-01)
The initial shear strength and changes in flexural strength of self-reinforced, absorbable polyglycolide (PGA) composite rods, submerged in distilled water (at 37 degrees C) for a period of 6 wk, were investigated. The recently developed self-reinforced absorbable material consists of
S B Pai
Mutation research, 119(3), 289-291 (1983-03-01)
Fenaminosulf (p-dimethylaminobenzenediazo sodium sulfonate, CAS registry No. 140-56-7) which is an active ingredient in several commercial fungicides was reported to be mutagenic in Salmonella typhimurium (McCann et al., 1975), Bacillus subtilis (Kada et al., 1974) and shown to cause chromosome
M Shun et al.
Zhong yao cai = Zhongyaocai = Journal of Chinese medicinal materials, 20(2), 56-58 (1997-02-01)
This paper reports the effect on nymexazol protecting the Rhizoctonia solani of Panax ginseng. The protective effect is in 80-95%. The drug can substitute quintozene and fenaminosulf in production.
Z Li et al.
Zhong yao cai = Zhongyaocai = Journal of Chinese medicinal materials, 21(4), 163-166 (2003-02-06)
Chemical Control tests of pot, plot and field for Panax notoginseng root rot were conducted during 1995-1996. The results indicated that the chemical control is a effect measure to control rapidly occurring and spreading of Panax notoginseng root rot. It

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service