Skip to Content
Merck
All Photos(2)

Key Documents

45469

Supelco

EPTC

PESTANAL®, analytical standard

Synonym(s):

S-Ethyl-N,N-dipropylthiocarbamate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H19NOS
CAS Number:
Molecular Weight:
189.32
Beilstein:
1762751
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

CCCN(CCC)C(=O)SCC

InChI

1S/C9H19NOS/c1-4-7-10(8-5-2)9(11)12-6-3/h4-8H2,1-3H3

InChI key

GUVLYNGULCJVDO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

230.0 °F

Flash Point(C)

110 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Vidal
Chemosphere, 36(12), 2593-2606 (1998-05-07)
Photocatalytic processes in the presence of titanium dioxide provide an interesting route to destroy hazardous organic contaminants, being operational in the UV-A domain with a potential use of solar radiation. In this paper, some specific contaminant classes of interest such
Dana M van Bemmel et al.
Environmental health perspectives, 116(11), 1541-1546 (2008-12-06)
The Agricultural Health Study (AHS) is a prospective cohort study of licensed pesticide applicators from Iowa and North Carolina enrolled between 1993 and 1997. EPTC (S-ethyl-N,N-dipropylthiocarbamate) is a thiocarbamate herbicide used in every region of the United States. The U.S.
I Nagy et al.
Applied and environmental microbiology, 61(5), 2056-2060 (1995-05-01)
During atrazine degradation by Rhodococcus sp. strain N186/21, N-dealkylated metabolites and an hydroxyisopropyl derivative are produced. The cytochrome P-450 system that is involved in degradation of thiocarbamate herbicides by strain N186/21 (I. Nagy, G. Schoofs, F. Compernolle, P. Proost, J.
I Nagy et al.
Journal of bacteriology, 177(3), 676-687 (1995-02-01)
Determination of the N-terminal sequences of two EPTC (S-ethyl dipropylcarbamothioate)-induced proteins from thiocarbamate-degrading Rhodococcus sp. strain NI86/21 resolved by two-dimensional electrophoresis enabled the localization of the respective structural genes on two distinct DNA fragments. One of these strongly induced proteins
Comparison of the persistence of EPTC, metribuzin, and propanil in Saskatchewan field soils.
A E Smith et al.
Bulletin of environmental contamination and toxicology, 29(2), 243-247 (1982-08-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service