Skip to Content
Merck
All Photos(1)

Key Documents

45357

Supelco

Bromopropylate

PESTANAL®, analytical standard

Synonym(s):

Isopropyl 4,4′-dibromobenzilate, Phenisobromolate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H16Br2O3
CAS Number:
Molecular Weight:
428.12
Beilstein:
2152560
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

CC(C)OC(=O)C(O)(c1ccc(Br)cc1)c2ccc(Br)cc2

InChI

1S/C17H16Br2O3/c1-11(2)22-16(20)17(21,12-3-7-14(18)8-4-12)13-5-9-15(19)10-6-13/h3-11,21H,1-2H3

InChI key

FOANIXZHAMJWOI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marc Tienstra et al.
Journal of AOAC International, 101(2), 342-351 (2017-11-17)
A method for residue analysis of pesticides and polychlorinated biphenyls in cereals and feed ingredients based on QuEChERS extraction, programmed temperature vaporizer large-volume injection, and GC with electron ionization (EI) quadrupole Orbitrap full-scan high-resolution MS (60 000 full width at half-maximum
Yu-Wen Huang et al.
Journal of chromatography. A, 1564, 51-58 (2018-06-16)
A dumbbell-shaped magnetic stir-bar with sublimate sorbent was prepared for the stir bar sorptive extraction (SBSE) of pesticides in an aqueous sample prior to gas chromatography-micro-electron capture detection (GC-μECD). Cyclododecane (CDD) was coated onto a magnetic stir-bar surface as a
M A Fernandez Muiño et al.
Journal of food protection, 60(1), 78-80 (1997-01-01)
Honey samples (101) from Galicia (N.W. Spain) were analyzed by gas chromatography (electron capture and flame ionization) for the presence of acaricides (amitraz, bromopropylate, coumaphos, and fluvalinate). Seventy-three samples were free from detectable residues. Bromopropylate residues were found in 16
Treatment of bee colonies with isopropyl-4,4-dibromo-benzilate against varroa disease and acarine disease.
W Ritter et al.
Zentralblatt fur Veterinarmedizin. Reihe B. Journal of veterinary medicine. Series B, 30(4), 266-273 (1983-05-01)
Ronnie Juraske et al.
Chemosphere, 70(10), 1748-1755 (2007-10-09)
Degradation half-lives in/on vegetation are needed in environmental risk assessment of pesticides, but these data are often not available for most active ingredients. To address this, we first correlated experimental soil degradation half-life data of 41 pesticides obtained from the

Protocols

Test your food for fipronil contamination using our analytical standards, certified reference materials, solvents, and columns for analysis.

Test your food for fipronil contamination using our analytical standards, certified reference materials, solvents, and columns for analysis.

Test your food for fipronil contamination using our analytical standards, certified reference materials, solvents, and columns for analysis.

Test your food for fipronil contamination using our analytical standards, certified reference materials, solvents, and columns for analysis.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service