Skip to Content
Merck
All Photos(1)

Key Documents

44964

Supelco

Density Standard 692 kg/m3

H&D Fitzgerald Ltd. Quality

Synonym(s):

2,2,4-Trimethylpentane, Isooctane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHCH2C(CH3)3
CAS Number:
Molecular Weight:
114.23
Beilstein:
1696876
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

H&D Fitzgerald Ltd. Quality
analytical standard

vapor density

3.9 (vs air)

vapor pressure

41 mmHg ( 21 °C)

autoignition temp.

745 °F

shelf life

limited shelf life, expiry date on the label

expl. lim.

6 %

manufacturer/tradename

H&D Fitzgerald Ltd.

refractive index

n20/D 1.391 (lit.)

bp

98-99 °C (lit.)

mp

−107 °C (lit.)

density

0.692 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
environmental
flavors and fragrances
food and beverages
industrial qc
personal care
petroleum
pharmaceutical (small molecule)

format

mixture

SMILES string

CC(C)CC(C)(C)C

InChI

1S/C8H18/c1-7(2)6-8(3,4)5/h7H,6H2,1-5H3

InChI key

NHTMVDHEPJAVLT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

This density CRM is used for the calibration of all types of laboratory density meters in the test temperature range of 10 to 25 °C.

Features and Benefits

  • Manufactured and verified in anISO 17025 accredited laboratory, as per UKAS
  • Carries an associated uncertaintyof +/- 0.010 Kg/m3
  • Provided in a sealed glassampoule of 10 mL to assure safe delivery
  • Accompanied with a certificate of analysis (CoA)having information on the expiry date for an effective audit

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

10.4 °F - closed cup

Flash Point(C)

-12 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kyung Min Park et al.
Food chemistry, 138(2-3), 733-738 (2013-02-16)
In this study, we investigated the possibility of reversed micelles, widely used as an enzyme reactor for lipases, for the determination of lipoxygenase activity. Although it is rapid and simple, reversed micelles have some limitations, such as interference by UV-absorbing
Yufeng Zhang et al.
Journal of chromatography. A, 1252, 67-73 (2012-07-10)
In this study, a polyethylene Pasteur pipette-based ultrasound-assisted emulsification microextraction (USAEME) applying low-density organic solvent was successfully developed for the extraction of trace levels of organochlorine pesticides (OCPs) in water samples and followed by gas chromatography-mass spectrometry (GC-MS) analysis. In
Swarna Bansal et al.
Journal of hazardous materials, 190(1-3), 45-50 (2011-04-05)
Separation of secondary emulsions of dispersed droplet size less than 10 μm, by means of fibrous medium is a very complex but important process. The study investigates the influence of thin fibrous filter properties, i.e. surface energy, pore size and
Luciane Batistella et al.
Bioprocess and biosystems engineering, 35(3), 351-358 (2011-07-23)
Both stability and catalytic activity of two commercial immobilized lipases were investigated in the presence of different organic solvents in ultrasound-assisted system. In a general way, for Novozym 435, the use of ethanol as solvent led to a loss of
Koichi Sawada et al.
Journal of bioscience and bioengineering, 114(2), 204-208 (2012-05-19)
Electrospun polyvinyl alcohol (PVA) nanofibrous non-woven fabrics have been widely used for cell and enzyme immobilization. Enhancement of the productivity of the material will further enlarge the versatility of them. In this study, a mixture of water and N,N-dimethylformamide (DMF)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service