Skip to Content
Merck
All Photos(1)

Key Documents

4465661

iDQuant Standards Kit for Pesticide

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
77101502
NACRES:
NA.24
Pricing and availability is not currently available.

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture

format

multi-component solution

shipped in

dry ice

storage temp.

−20°C

General description

The iDQuant Standards Kit for Pesticide Analysis contains 204 common pesticides and can be used for quantifying and identifying pesticides in any sample matrix.  The mix contains 10 mixes of about 20 pesticides each at initial concentrations of 100 μg/mL in 100% acetonitrile.  It is also stable for 1 year when stored at -20 ?C.  Certificate of Analysis is provided by manufacturer who is accredited to ISO Guide 34, ISO/IEC 17025 and ISO 9001:2008.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Analysis Note

To view the Bulletin for the iDQuant Standards Kit for Pesticide Analysis, please visit this Bulletin link.

To view an Application Note for the iDQuant Standards Kit for Pesticide Analysis, please visit this Application Note link.

Legal Information

iDQuant is a trademark of AB Sciex Pte. Ltd.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

Flash Point(F)

35.6 °F - closed cup

Flash Point(C)

2.0 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service