Skip to Content
Merck

44462

AmberChrom 50WX2 Ion Exchange Resin

sodium form, 100-200 mesh

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

CAS Number:
UNSPSC Code:
23151817
NACRES:
SB.52
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

AmberChrom 50WX2, Na+-form, 100-200 mesh

form

beads

crosslinking

2 % cross-linked

technique(s)

LPLC: suitable

loss

~75% loss on drying, 110 °C

matrix

styrene-divinylbenzene

matrix active group

sulfonic acid functional group

particle size

100-200 mesh

capacity

0.6 meq/mL

separation technique

cation exchange

SMILES string

[Na+].[C-H2]Cc3c(cccc3)C=C.c2(c(cccc2)C=C)C=C.c1(ccccc1)C=C

InChI

1S/C10H11.C10H10.C8H8.Na/c2*1-3-9-7-5-6-8-10(9)4-2;1-2-8-6-4-3-5-7-8;/h3,5-8H,1-2,4H2;3-8H,1-2H2;2-7H,1H2;/q-1;;;+1

InChI key

NEQWAZPDMBONCG-UHFFFAOYSA-N

Legal Information

Amberchrom is a trademark of DuPont de Nemours, Inc.


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library