Skip to Content
Merck

44324

AmberChrom 1X8 Ion Exchange Resin

chloride form, 20-50 mesh

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
(C10H12 · C10H10 · C8H8 · C3H9N)X
CAS Number:
UNSPSC Code:
23151817
NACRES:
SB.52
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

AmberChrom 1X8 chloride form, chloride form, 20-50 mesh

Quality Segment

description

cross-linkage 8%, strongly basic

form

beads

parameter

66 °C OH- form max. temp.

technique(s)

LPLC: suitable

loss

~50% loss on drying, 110 °C

matrix

styrene-divinylbenzene (gel)

matrix active group

quaternary ammonium

particle size

20-50 mesh

operating pH

0-14

capacity

1.2 meq/mL by wetted bed volume

separation technique

anion exchange

SMILES string

N(C)(C)C.C(C)c3c(cccc3)C=C.c2(c(cccc2)C=C)C=C.c1(ccccc1)C=C

InChI

1S/C10H12.C10H10.C8H8.C3H9N/c2*1-3-9-7-5-6-8-10(9)4-2;1-2-8-6-4-3-5-7-8;1-4(2)3/h3,5-8H,1,4H2,2H3;3-8H,1-2H2;2-7H,1H2;1-3H3

InChI key

HADXLRSCRPYPJJ-UHFFFAOYSA-N

General description

"p.a." grade resins have been cleaned to remove residual metals, and are purified for analytical use.
Dowex 1X8 is a strongly basic anion exchange resin which is used as an absorbent in solid phase extraction.

Application

Dowex 1X8 chloride form was used:
  • to separate the 3H2O from D[2-3H] glucose using Dowex columns
  • in a assay to determine the pantothenate kinase using Dowex column
  • to separate inositol phosphates (IPs) from myo-[3H]inositol by Dowex chromatography
Strongly basic (type I) anion exchange resin for fine chemical processing; meets requirements of FDA Food Additive Regulation 21 CFR 173.25.

Legal Information

Amberchrom is a trademark of DuPont de Nemours, Inc.

Disclaimer

may contain black particles


Still not finding the right product?

Explore all of our products under AmberChrom 1X8 chloride form


Storage Class

11 - Combustible Solids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library