Skip to Content
Merck
All Photos(2)

Documents

44178

Supelco

1-Phenyldodecane

analytical standard

Synonym(s):

Dodecylbenzene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5(CH2)11CH3
CAS Number:
Molecular Weight:
246.43
Beilstein:
1909107
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥99.5% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.482 (lit.)
n20/D 1.482

bp

185-188 °C/15 mmHg (lit.)
331 °C (lit.)

mp

3 °C (lit.)

density

0.856 g/mL at 25 °C (lit.)

application(s)

environmental

format

neat

SMILES string

CCCCCCCCCCCCc1ccccc1

InChI

1S/C18H30/c1-2-3-4-5-6-7-8-9-10-12-15-18-16-13-11-14-17-18/h11,13-14,16-17H,2-10,12,15H2,1H3

InChI key

KWKXNDCHNDYVRT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

228.2 °F - closed cup

Flash Point(C)

109 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Studies of the carcinogenesis and tumorigenesis of skin applications of dodecylbenzene on hairless mice.
C B Assogne
British journal of industrial medicine, 47(5), 356-358 (1990-05-01)
Ding-xi Cheng et al.
Guang pu xue yu guang pu fen xi = Guang pu, 27(1), 104-107 (2007-03-30)
There is a self-polymerization equilibrium system between the cation of the fluorescent dye-safranine T and its dimer. In the presence of appropriate amounts of anionic surfactant SDBS, the above mentioned equilibrium can be adjusted by a quantitative addition of protein
J Campos-García et al.
Applied and environmental microbiology, 65(8), 3730-3734 (1999-07-31)
Pseudomonas aeruginosa W51D is able to grow by using branched-chain dodecylbenzene sulfonates (B-DBS) or the terpenic alcohol citronellol as a sole source of carbon. A mutant derived from this strain (W51M1) is unable to degrade citronellol but still grows on
C Ruffo et al.
Ecotoxicology and environmental safety, 8(3), 275-279 (1984-06-01)
Pyridine and nitrilotriacetic acid sodium salt as models of soluble compounds and linear dodecylbenzene, branched dodecylbenzene, and stearic acid as models of insoluble compounds were compared for their biodegradability in three tests: the ODCE modified test and the Sturm and
O H Iversen
British journal of industrial medicine, 46(9), 608-616 (1989-09-01)
Dodecylbenzene in various concentrations, dissolved in acetone to a final volume of 50 microliters, was applied twice a week for 78 weeks to the back skin of hairless mice, with or without pretreatment with 51.2 micrograms 9,10-dimethyl-1,2-benzanthracene (DMBA). A negative

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service