Skip to Content
Merck
All Photos(1)

Key Documents

439126

Sigma-Aldrich

Acetone

suitable for HPLC, ≥99.9%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COCH3
CAS Number:
Molecular Weight:
58.08
Beilstein:
635680
EC Number:
MDL number:
UNSPSC Code:
12190000
PubChem Substance ID:
NACRES:
NA.02
grade:
HPLC grade
technique(s):
HPLC: suitable
bp:
56 °C/760 mmHg (lit.)
vapor pressure:
184 mmHg ( 20 °C)
Pricing and availability is not currently available.

grade

HPLC grade

Quality Level

vapor density

2 (vs air)

vapor pressure

184 mmHg ( 20 °C)

Assay

≥99.9%

form

liquid

expl. lim.

13.2 %

technique(s)

HPLC: suitable

impurities

≤0.5% (water)

evapn. residue

<0.0002%

color

clear colorless

refractive index

n20/D 1.359 (lit.)

bp

56 °C/760 mmHg (lit.)

mp

−94 °C (lit.)

solubility

water: miscible

density

0.791 g/mL at 25 °C (lit.)

λ

H2O reference

UV absorption

λ: 330 nm Amax: 1.00
λ: 340 nm Amax: 0.10
λ: 350 nm Amax: 0.02
λ: 400 nm Amax: 0.01

suitability

suitable (pesticide analysis)

format

neat

SMILES string

CC(C)=O

InChI

1S/C3H6O/c1-3(2)4/h1-2H3

InChI key

CSCPPACGZOOCGX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Acetone is a ketone. Mixtures of TiO2 (P25) with rare earth oxides (La2O3 or Y2O3) calcined at 700°C or 650°C has been reported to efficiently photocatalyze the oxidation of acetone.[1] Its aldol condensation in vapor-phase has been investigated over MgO promoted with alkali (Li, Na, K and Cs) or alkaline earth (Ca, Sr and Ba) metal ions.[2] It undergoes Aldol condensation in the presence of Mg-Al layered double hydroxides (LDH) as catalysts and Cl- and/or CO32- as compensating anions to afford diacetone alcohol and mesityl oxide.[3] Enantioselective Aldol condensation of acetone with various isatins in the presence of dipeptides (catalyst) has been described.[4]

Application

Acetone was used in the affinity purification of modified proteins using streptavidin beads for the mass spectrometric (MS) identification of poly(ADP-ribose) polymerases (PARP) substrate proteins.[5]
It may be used in the following studies:
  • NMR spectroscopic evaluation of glucose metabolism in biological samples (blood).[6]
  • Preparation of 13wt% amorphous poly (lactic acid) (PLA) solution, which was required for the preparation of electrospun PLA membranes.[7]
  • As a precursor for the synthesis of methyl isobutyl ketone (MIBK) in the presence of sulfonated graphene oxide-Pd/cordierite catalyst.[8]
  • Synthesis of (4-hydroxymethyl-2,2-dimethyl-1,3-dioxolane), a solketal from glycerol using supercritical fluids (SCF) technology.[9]
It may be used in the synthesis of the following acetone hydrazones:
  • 1-isopropylidene-2-methylhydrazine[10]
  • 1-isopropylidene-2-hydroxyethylhydrazine[10]
  • 1-isopropylidene-2-formylhydrazine[10]
Acetone′s luminesence intensity is dependent upon the solution components . The absorption of UV light by acetone, results in its photolysis and the production of radials .

Packaging

M-Bottle for Solvents
As a global leader in lab reagents, we are constantly looking for new ways to optimize the safety of our products. The newly developed 4L solvent bottle design features advanced sealing technology that eliminates leaks to make the handling of solvents safer and more convenient than ever before.
See all the new features here!

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

1.4 °F - closed cup

Flash Point(C)

-17.0 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 3

1 of 3

Ketalization of glycerol to solketal in supercritical acetone.
Royon D, et al.
Journal of Supercritical Fluids, 58(1), 88-92 (2011)
Sulfonated graphene oxide supported Pd bifunctional catalyst for one-pot synthesis of methyl isobutyl ketone from acetone with high conversion and selectivity.
Liu M, et al.
J. Mol. Catal. A: Chem., 408, 85-90 (2015)
Saskia Kley et al.
American journal of physiology. Regulatory, integrative and comparative physiology, 296(4), R936-R943 (2009-02-06)
Obesity is a risk factor for type 2 diabetes in cats. The risk of developing diabetes is severalfold greater for male cats than for females, even after having been neutered early in life. The purpose of this study was to
Development of eco-efficient micro-porous membranes via electrospinning and annealing of poly (lactic acid).
Li L, et al.
Journal of Membrane Science, 436, 57-67 (2013)
Hong Jiang et al.
Current protocols in chemical biology, 4(1), 19-34 (2012-11-29)
Poly(ADP-ribose) polymerases (PARPs) play important roles in various biological processes, including DNA repair, transcriptional regulation, mitosis, and RNA processing. PARP inhibitors are in clinical trials for treating human cancers. Understanding the biological function of PARPs will be important to fully

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service