Skip to Content
Merck

436615

Amberlite HPR650 Ion Exchange Resin

hydrogen form, 25-30 mesh, cationic

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

CAS Number:
UNSPSC Code:
23151817
NACRES:
SB.52
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

AmberLite HPR650 H, Cation Exchange Resin

Quality Segment

description

cross-linkage 8%, formerly Dowex Monosphere 650 C hydrogen form

form

beads

parameter

150 °C max. temp.

moisture

46-51%

technique(s)

LPLC: suitable

matrix

styrene-divinylbenzene (gel)

matrix active group

sulfonic acid functional group

particle size

21-25 mesh, 600-700 μm

operating pH

0-14

capacity

2.0 meq/mL by wetted bed volume

bulk density

690 g/L

separation technique

cation exchange

SMILES string

C(C)c3c(cccc3)C=C.c2(c(cccc2)C=C)C=C.c1(ccccc1)C=C

InChI

1S/C10H12.C10H10.C8H8/c2*1-3-9-7-5-6-8-10(9)4-2;1-2-8-6-4-3-5-7-8/h3,5-8H,1,4H2,2H3;3-8H,1-2H2;2-7H,1H2

InChI key

NWUYHJFMYQTDRP-UHFFFAOYSA-N

General description

AmberLite HPR650 H is a strong basic cation exchange gel resin with sulfonic acid as functional group.AmberLite HPR650 H is a heterogeneous catalyst.

Application

AmberLiteHPR650 H packed column was used in flow based ammonia gas analyzer.
Strongly acidic cation exchange resin for mixed-bed applications. Commonly used for condensate polishing and sugar applications. This resin meets requirements of FDA Food Additive Regulation 21 CFR 173.25.

Features and Benefits

Strongly acidic cation.

Other Notes

This ion-exchange resin has not been specially processed nor cleaned. We suggest that it be treated to a suitable preliminary elution and wash.

Legal Information

Amberlite is a trademark of DuPont de Nemours, Inc.


pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Eye Irrit. 2

Storage Class

11 - Combustible Solids

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library


Related Content


Kinetic Study of Esterification of Acetic Acid with n-butanol and isobutanol Catalyzed by Ion Exchange Resin
Toor, Amrit Pal, et al.
Bulletin of Chemical Reaction Engineering and Catalysis, 6 (2011)
Optimization of the reverse osmosis seawater demineralization technologies for a power producing industry
Fendri, Fredg, Tatyana Mitchenko, and Zakhar Maletskyi
Desalination and Water Treatment, 25, 84-90 (2011)
Shin-Ichi Ohira et al.
Talanta, 116, 527-534 (2013-10-24)
A robust and fully automated indoor ammonia gas monitoring system with an open channel scrubber (OCS) was developed. The sample gas channel dimensions, hydrophilic surface treatment to produce a thin absorbing solution layer, and solution flow rate of the OCS



Global Trade Item Number

SKUGTIN
436615-1KG04061826750186
436615-250G04061838513892