Skip to Content
Merck
All Photos(1)

Key Documents

431400

Supelco

Phosphomolybdic acid hydrate

≥99.99% trace metals basis

Synonym(s):

Molybdophosphoric acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H3[P(Mo3O10)4] · xH2O
CAS Number:
Molecular Weight:
1825.25 (anhydrous basis)
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.21

Quality Level

Assay

≥99.99% trace metals basis

form

crystals

reaction suitability

reagent type: oxidant

impurities

≤0.01% insolubles
<100 ppm total metallic impurities

density

1.62 g/mL at 25 °C (lit.)

anion traces

chloride (Cl-): ≤0.02%
sulfate (SO42-): ≤0.025%

cation traces

Ca: ≤0.02%
Fe: ≤0.005%
NH4+: ≤0.01%
heavy metals: ≤0.005%

SMILES string

O.O=[Mo](=O)=O.O=[Mo](=O)=O.O=[Mo](=O)=O.O=[Mo](=O)=O.O=[Mo](=O)=O.O=[Mo](=O)=O.O=[Mo](=O)=O.O=[Mo](=O)=O.O=[Mo](=O)=O.O=[Mo](=O)=O.O=[Mo](=O)=O.O=[Mo](=O)=O.OP(O)(O)=O

InChI

1S/12Mo.H3O4P.H2O.36O/c;;;;;;;;;;;;1-5(2,3)4;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;/h;;;;;;;;;;;;(H3,1,2,3,4);1H2;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;

InChI key

PDDXOPNEMCREGN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Phosphomolybdic acid hydrate is a heteropolyacid hydrate. It participates in the synthesis of molybdenum disulfide (MoS2)-reduced graphene oxide (RGO) composites. Phosphomolybdic acid is one of the components of the Goldner′s trichrome stain used for tissue staining.

Application

Employed in preparation of a hybrid organic-inorganic, electroactive material with potential application as a cation-insertion electrode in batteries.
Reactant for preparation of:
  • Catalysts for the nitration of aromatic compounds
  • Highly dispersed MoOx/CNTs (carbon nanotubes) as a support for Pt catalysts
  • Reduced molybdophosphate mixed-valence Keggin clusters
  • Polyoxometalate (POM) hybrid materials based on Keggin polyanion building blocks
  • Luminescent Langmuir-Blodgett films
  • Anchored Wilkinson catalyst
A precursor to hybrid organic-inorganic based molecular batteries with applications as cation-insertion electrodes.

Features and Benefits

Meets A.C.S. reagent specifications.

Pictograms

Flame over circleCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Ox. Sol. 3 - Skin Corr. 1B

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Liu, L.; et al.
Journal of Organometallic Chemistry, 696, 1319-1319 (2011)
Tanielyan, S. K.; et al.
ACS Catalysis, 1, 159-159 (2011)
Liu, H.-Y.; et al.
Crystal Growth & Design, 11, 1786-1786 (2011)
Tomoyuki Akutagawa et al.
Inorganic chemistry, 50(14), 6711-6718 (2011-06-16)
Hydrogen-bonded assemblies of the two-electron reduced mixed-valence Keggin clusters [PMo(12)O(40)](5-) and [SiMo(12)O(40)](6-) were obtained by the one-pot electron-transfer reactions between p-phenylenediamine (PPD) or 2,3,5,6-tetramethyl-PPD (TMPPD) (donors) and H(+)(3)[PMo(12)O(40)](3-) or H(+)(4)[SiMo(12)O(40)](4-) (acceptors) in CH(3)CN. The redox states of the [PMo(12)O(40)](5-) and
Zhi Ming Cui et al.
Chemical communications (Cambridge, England), 47(29), 8418-8420 (2011-06-28)
HPMo self-assembly on carbon nanotubes followed by decomposition is used to fabricate highly dispersed MoO(x)/CNTs as a support for high performance of a Pt catalyst towards methanol oxidation.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service