Skip to Content
Merck

429554

Acetaldehyde-2,4-dinitrophenylhydrazone

analytical standard

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
CH3CH=NNHC6H3(NO2)2
CAS Number:
Molecular Weight:
224.17
UNSPSC Code:
12171500
eCl@ss:
39021102
PubChem Substance ID:
NACRES:
NA.47
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

analytical standard

Quality Segment

assay

99%

form

powder or crystals

technique(s)

titration: suitable

mp

165-168 °C (lit.)

application(s)

diagnostic assay manufacturing
environmental
hematology
histology

storage temp.

room temp

SMILES string

C\C=N\Nc1ccc(cc1[N+]([O-])=O)[N+]([O-])=O

InChI

1S/C8H8N4O4/c1-2-9-10-7-4-3-6(11(13)14)5-8(7)12(15)16/h2-5,10H,1H3/b9-2+

InChI key

ONBOQRNOMHHDFB-XNWCZRBMSA-N



Still not finding the right product?


pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library