Skip to Content
Merck

428663

AmberChrom 50WX4 Ion Exchange Resin

hydrogen form, 50-100 mesh

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
(C10H12 · C10H10 · C8H8)x
CAS Number:
UNSPSC Code:
23151817
NACRES:
SB.52
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

AmberChrom 50WX4 hydrogen form, hydrogen form, 50-100 mesh

description

cross-linkage 4%

Quality Segment

form

beads

moisture

64-72%

technique(s)

LPLC: suitable

matrix

styrene-divinylbenzene (gel)

matrix active group

sulfonic acid functional group

particle size

50-100 mesh

capacity

1.1 meq/mL by wetted bed volume

separation technique

cation exchange

SMILES string

C(C)c3c(cccc3)C=C.c2(c(cccc2)C=C)C=C.c1(ccccc1)C=C

InChI

1S/C10H12.C10H10.C8H8/c2*1-3-9-7-5-6-8-10(9)4-2;1-2-8-6-4-3-5-7-8/h3,5-8H,1,4H2,2H3;3-8H,1-2H2;2-7H,1H2

InChI key

NWUYHJFMYQTDRP-UHFFFAOYSA-N

General description

Dowex 50WX4 hydrogen form is a nonmacroporous gelform bead resin. Dowex 50WX4 hydrogen form can be used as a column packing material which is used in anion exchange chromatography.

Application

Dowex 50WX4 hydrogen form was used:
  • to investigate the effect of particle size on drug loading and release profiles
  • for conversion of D,L-glyceraldehyde 3-phosphate diethyl acetal (barium salt) on treatment with boiling water to free aldehyde D,L-glyceraldehyde 3-phosphate
Strongly acidic cation exchange resin that meets requirements of FDA Food Additive Regulation 21 CFR 173.25

Other Notes

This strongly acidic cation exchange resin has not been specially processed nor cleaned. We suggest that it be subjected to a suitable preliminary elution and wash.

Legal Information

Amberchrom is a trademark of DuPont de Nemours, Inc.


Still not finding the right product?

Explore all of our products under AmberChrom 50WX4 hydrogen form


pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Eye Irrit. 2

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library