Skip to Content
Merck
All Photos(1)

Key Documents

41605

Supelco

DL-Mandelic acid

analytical standard

Synonym(s):

(±)-α-Hydroxyphenylacetic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH(OH)COOH
CAS Number:
Molecular Weight:
152.15
Beilstein:
510011
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

Quality Level

grade

analytical standard

Assay

≥98.5% (HPLC)

optical activity

[α]/D 0±1°, c = 5 in H2O

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

cleaning products
cosmetics
food and beverages
forensics and toxicology
personal care
veterinary

format

neat

SMILES string

OC(C(O)=O)c1ccccc1

InChI

1S/C8H8O3/c9-7(8(10)11)6-4-2-1-3-5-6/h1-5,7,9H,(H,10,11)

InChI key

IWYDHOAUDWTVEP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

DL-Mandelic acid is an aromatic hydrocarbon metabolite of styrene.[1][2]

Application

DL-Mandelic acid may be used as an analytical reference standard for the determination of DL-mandelic acid in:
  • Human urine samples by high performance liquid chromatography (HPLC) equipped with ultraviolet (UV) detector.[1]
  • Rat urine samples by gas chromatography-mass spectrometry (GC-MS) with selected ion monitoring (SIM) detection.[2]

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Simultaneous determination of urinary hippuric acid, o-, m-and p-methylhippuric acids, mandelic acid and phenylglyoxylic acid for biomonitoring of volatile organic compounds by gas chromatography-mass spectrometry.
Ohashi Y, et al.
Analytica Chimica Acta, 566(2), 167-171 (2006)
H Crespi et al.
The Journal of investigative dermatology, 78(6), 493-497 (1982-06-01)
The purpose of this study was to evaluate the in vivo state of both branches of the autonomic nervous system in children with chronic atopic dermatitis. In 15 patients, age 4 to 11, the following parameters were analyzed: (1) basal
P Vodicka et al.
Carcinogenesis, 16(7), 1473-1481 (1995-07-01)
Occupational exposure to styrene was studied in nine workers of a hand lamination plant in Bohemia. Personal dosimeters were used to monitor the styrene workplace exposure, and the levels of styrene in blood and mandelic acid in urine were measured.
Ludmila Krivánková et al.
Electrophoresis, 24(3), 505-517 (2003-02-06)
Effects originating from the variability of the sample matrix can be efficiently eliminated when the separation conditions are selected so that compounds of like charge with high concentration referred to as macrocomponents are embodied into the system of transient isotachophoresis.
A Kimura et al.
Pediatric research, 45(4 Pt 1), 603-609 (1999-04-15)
Unusual bile acids, such as unsaturated ketonic and 7beta-hydroxylated bile acids, have been detected in urine early in life. To elucidate the normal profiles of usual and unusual urinary bile acids in the neonatal and pediatric periods, we measured the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service