415530PLATER
Blue dye for platers
for use with easySpiral (platers), pkg of 125 mL
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41122409
NACRES:
NB.24
packaging
pkg of 125 mL
application(s)
cosmetics
food and beverages
pharmaceutical
compatibility
for use with easySpiral (platers)
General description
Color dye for Spiral tests (125 mL) for easySpiral® plater
Application
Culture media plating
Legal Information
easySpiral is a registered trademark of InterLab
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service