Skip to Content
Merck

41206

Neosaxitoxin-15N7 solution

10 μg/g in hydrochloric acid (nominal concentration), certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Synonym(s):

neo-Saxitoxin-15N7 solution, NeoSTX-15N7

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C10H1715N7O5
Molecular Weight:
322.24
MDL number:
UNSPSC Code:
41116107
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

grade

certified reference material, TraceCERT®

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

concentration

10 μg/g in hydrochloric acid (nominal concentration)

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

single component solution

storage temp.

−20°C

SMILES string

[H][C@@]12[C@]3([15NH]C([15NH]2)=[15NH])[15N](CCC3(O)O)C([15N](O)[C@H]1COC([15NH2])=O)=[15NH]

InChI key

PPEKGEBBBBNZKS-FXWZNOLFSA-N

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to the SI through a primary reference material from a NMI.
Certified content incl. uncertainty and expiry date are stated on the enclosed certificate.

Paralytic Shellfish Toxin PST for the analysis of marine toxins in seafood.

Check out our complete range of marine toxins reference materials

This is the 13C isotope-labelled analog of Neosaxitoxin

Analysis Note

10 μg/g in 3 mmol/l HCl in water

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany


Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library